RAB0471
Rat TIMP-1 ELISA Kit
for cell and tissue lysates
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
UNSPSC Code:
41116158
NACRES:
NA.32
Recommended Products
species reactivity
rat
packaging
kit of 96 wells (12 strips x 8 wells)
technique(s)
ELISA: suitable
capture ELISA: suitable
input
sample type tissue lysate
sample type cell lysate
assay range
inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 4 pg/mL
standard curve range: 4.1-3000 pg/mL
detection method
colorimetric
shipped in
wet ice
storage temp.
−20°C
Gene Information
rat ... Timp1(116510)
Related Categories
General description
The Rat TIMP-1 ELISA (Enzyme-Linked Immunosorbent Assay) kit is an in vitro enzyme-linked immunosorbent assay for the quantitative measurement of Rat TIMP-1 cell lysate and tissue lysate.
Immunogen
Recombinant Rat TIMP1
Application
For research use only. Not for use in diagnostic procedures.
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)
Other Notes
A sample Certificate of Analysis is available for this product.
Please type the word sample in the text box provided for lot number.
Please type the word sample in the text box provided for lot number.
Kit Components Also Available Separately
Product No.
Description
SDS
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Met. Corr. 1
Storage Class Code
8A - Combustible corrosive hazardous materials
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Hirokuni Akahori et al.
Journal of hypertension, 32(7), 1534-1541 (2014-04-25)
Clinical studies have suggested the beneficial effects of statin therapy on diastolic heart failure. However, the mechanism of the beneficial effects of statin on diastolic heart failure remains unknown. We examined the effect of atorvastatin on the cardiac function of
Nejat Nizam et al.
Inflammation, 37(5), 1771-1778 (2014-05-07)
Salivary, serum matrix metalloproteinase-8 (MMP-8), tissue inhibitor of matrix metalloproteinases-1 (TIMP-1), neutrophil elastase (NE), and myeloperoxidase (MPO) levels were investigated in generalized chronic periodontitis (GCP), generalized aggressive periodontitis (GAgP), and healthy groups. Whole-mouth clinical periodontal measurements were recorded. Salivary, serum
Harpreet Sidhu et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 34(30), 9867-9879 (2014-07-25)
Fmr1 knock-out (ko) mice display key features of fragile X syndrome (FXS), including delayed dendritic spine maturation and FXS-associated behaviors, such as poor socialization, obsessive-compulsive behavior, and hyperactivity. Here we provide conclusive evidence that matrix metalloproteinase-9 (MMP-9) is necessary to
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service