RAB0420
Human Serum Amyloid A ELISA Kit
for serum, plasma, cell culture supernatant and urine
Synonym(s):
SAA, Serum amyloid A-1 protein
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
UNSPSC Code:
41116158
NACRES:
NA.32
Recommended Products
species reactivity
human
packaging
kit of 96 wells (12 strips x 8 wells)
technique(s)
ELISA: suitable
capture ELISA: suitable
input
sample type cell culture supernatant(s)
sample type urine
sample type plasma
sample type serum
assay range
inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 500 pg/mL
standard curve range: 0.41-300 ng/mL
detection method
colorimetric
shipped in
wet ice
storage temp.
−20°C
Gene Information
human ... SAA1(6288)
General description
Serum amyloid A1 (SAA1) is an acute response protein. It is synthesized by the liver, during infection. SAA1 is expressed in the amnion and chorion of human fetal membranes. It is located on human chromosome chromosome 11. SAA1 encodes for the inducible forms of serum amyloid A (SAA) proteins.
The Human SAA (Serum Amyloid A) ELISA (Enzyme- Linked Immunosorbent Assay) kit is an in vitro enzyme-linked immunosorbent assay for the quantitative measurement of human SAA in serum, plasma, cell culture supernatants and urine.
Immunogen
Recombinant Human SAA
Application
For research use only. Not for use in diagnostic procedures.
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)
Biochem/physiol Actions
Serum amyloid A1 (SAA1) induces the generation of proinflammatory cytokines like, IL-1β (interleukin 1 β), interleukin 6 (IL-6) and the uterotonic hormone proinflammatory cytokines and prostaglandin E2 (PGE2).
Other Notes
A sample Certificate of Analysis is available for this product.
Please type the word sample in the text box provided for lot number.
Please type the word sample in the text box provided for lot number.
Kit Components Also Available Separately
Product No.
Description
SDS
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Met. Corr. 1
Storage Class Code
8A - Combustible corrosive hazardous materials
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Induction of pro-inflammatory genes by serum amyloid A1 in human amnion fibroblasts
Li W, et al.
Scientific reports, 7(1), 693-693 (2017)
The human serum amyloid A protein (SAA) superfamily gene cluster: mapping to chromosome 11p15. 1 by physical and genetic linkage analysis
Sellar G C, et al.
Genomics, 19(2), 221-227 (1994)
Melissa G Works et al.
Journal of immunology (Baltimore, Md. : 1950), 193(7), 3278-3287 (2014-08-27)
Psoriasis is a chronic autoimmune disease affecting the skin and characterized by aberrant keratinocyte proliferation and function. Immune cells infiltrate the skin and release proinflammatory cytokines that play important roles in psoriasis. The Th17 network, including IL-23 and IL-22, has
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service