RAB0236
Mouse IGFBP-3 ELISA Kit
for serum, plasma and cell culture supernatant
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
UNSPSC Code:
41116158
NACRES:
NA.32
Recommended Products
species reactivity
mouse
packaging
kit of 96 wells (12 strips x 8 wells)
technique(s)
ELISA: suitable
capture ELISA: suitable
input
sample type cell culture supernatant(s)
sample type serum
sample type plasma
assay range
inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 70 pg/mL
standard curve range: 54.87-40000 pg/mL
detection method
colorimetric
shipped in
wet ice
storage temp.
−20°C
Gene Information
mouse ... Igfbp3(16009)
Related Categories
General description
The Mouse IGFBP-3 (Insulin-like growth factor binding protein 3) ELISA (Enzyme-Linked Immunosorbent Assay) kit is an in vitro enzyme-linked immunosorbent assay for the quantitative measurement of Mouse IGFBP-3 in serum, plasma, cell culture supernatants and urine.
Immunogen
Recombinant Mouse IGFBP3
Application
For research use only. Not for use in diagnostic procedures.
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)
Other Notes
A sample Certificate of Analysis is available for this product.
Please type the word sample in the text box provided for lot number.
Please type the word sample in the text box provided for lot number.
Kit Components Also Available Separately
Product No.
Description
SDS
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Met. Corr. 1
Storage Class Code
8A - Combustible corrosive hazardous materials
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Na Ri Kim et al.
American journal of physiology. Endocrinology and metabolism, 320(3), E415-E424 (2020-12-15)
Sex steroids are critical for skeletal development and maturation during puberty as well as for skeletal maintenance during adult life. However, the exact time during puberty when sex steroids have the highest impact as well as the ability of bone
Na Ri Kim et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 34(5), 7118-7126 (2020-04-03)
Sexually dimorphic bone structure emerges largely during puberty. Sex steroids are critical for peak bone mass acquisition in both genders. In particular, the biphasic effects of estrogens mediate the skeletal sexual dimorphism. However, so far the stimulatory vs inhibitory actions
Yan Wang et al.
Medical oncology (Northwood, London, England), 31(5), 921-921 (2014-03-29)
Growth factors in the brain are important to depression. We investigated the relationship between serum insulin-like growth factor (IGF-1) and IGF-binding protein-3 (IGFBP-3) concentration and risk of depression and the effect of psychological intervention on outcomes of high-grade glioma patients
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service