Skip to Content
Merck
All Photos(1)

Key Documents

P5739

Sigma-Aldrich

Placenta Growth Factor-2 from mouse

lyophilized powder, recombinant, expressed in baculovirus infected Sf21 cells, suitable for cell culture

Synonym(s):

PGF-2, PlGF-2

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352202
NACRES:
NA.77

biological source

mouse

Quality Level

recombinant

expressed in baculovirus infected Sf21 cells

Assay

>97% (SDS-PAGE)

form

lyophilized powder

mol wt

~38 kDa by SDS-PAGE

packaging

pkg of 10 μg

storage condition

avoid repeated freeze/thaw cycles

technique(s)

cell culture | mammalian: suitable

impurities

endotoxin, tested

UniProt accession no.

storage temp.

−20°C

Gene Information

mouse ... Pgf(18654)

Biochem/physiol Actions

Angiogenic factor of the VEGF family. Ligand for the VEGFR-1 (flt-1) receptor and for neuropilin-1. PlGF-2 differs from PlGF-1 and PlGF-3 in having a heparin binding domain.

Physical form

Lyophilized from a 0.2 μm filtered solution in 30% acetonitrile and 0.1% trifluoroacetic acid containing 0.5 mg bovine serum albumin.

Analysis Note

The biological activity is measured by its ability to bind to immobilized Flt1/Fc chimera in ELISA.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Skin Corr. 1A

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

VEGF-A stimulates blood vessel growth via endothelial cell proliferation, migration, and survival during embryonic development.

VEGF-A stimulates blood vessel growth via endothelial cell proliferation, migration, and survival during embryonic development.

VEGF-A stimulates blood vessel growth via endothelial cell proliferation, migration, and survival during embryonic development.

VEGF-A stimulates blood vessel growth via endothelial cell proliferation, migration, and survival during embryonic development.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service