OH30456413
Ohaus® Navigator® Portable Balance
Model NV621, weighing capacity 620 g, resolution: 0.1 g, AC/DC input 110 V, US 3-pin plug (North America)
All Photos(1)
About This Item
UNSPSC Code:
41111502
Recommended Products
material
metal weighing plate
feature
CSA approved
packaging
pack of 1 ea
manufacturer/tradename
Ohaus® 30456413
weighing capacity measuring range
620 g, resolution: 0.1 g
weighing pan W × L
14.4 cm × 19 cm
General description
OHAUS raises the bar in price-performance ratio with the newly-designed Navigator series which is perfect for general weighing applications in laboratory, industrial and education settings. Ruggedly constructed and multifunctional, the Navigator′s a powerful balance that can handle a diverse range of weighing applications with features such as fast stabilization time, overload protection at four times its rated capacity, and easy operation.
Application
Weighing, Parts Counting, Percent Weighing, Checkweighing
Features and Benefits
Menu and calibration lock switch, mechanical and software overload/underload protection, stability indicator, low battery indicator, auto shut-off, user-selectable print options, user-selectable communication settings
Legal Information
Navigator is a registered trademark of Ohaus Corp.
Ohaus is a registered trademark of Ohaus Corp.