Skip to Content
Merck
All Photos(1)

Key Documents

M6062

Greiner HLA Terasaki multiwell plates

60 well space saver design, 1 lid per plate

Synonym(s):

60 well microtest plate, HLA microplates, HLA microtiter plates, HLA multiwell plates, Greiner HLA Terasaki multiwell plates

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122107
NACRES:
NB.22
material:
clear polystyrene
flat clear wells
size:
60 wells
sterility:
non-sterile
binding type:
Tissue Culture (TC)-treated surface
Tissue Culture (TC)-treated surface
feature:
lid
Pricing and availability is not currently available.

material

clear polystyrene
flat clear wells

sterility

non-sterile

feature

lid

packaging

case of 1200 ea
pack of 150 ea

manufacturer/tradename

Greiner 659190

size

60 wells

well size

10 μL

well volume

10 μL

binding type

Tissue Culture (TC)-treated surface
Tissue Culture (TC)-treated surface

Looking for similar products? Visit Product Comparison Guide

General description

HLA Microtest plates (Terasaki) from Greiner. Designed for tissue typing techniques to ensure smooth spreading of sera and cells across the flat base of the well. The surface of each well gives a microscopically precise and clear picture. All plates are stackable.

Legal Information


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service