Key Documents
LIDFA45211S-EP
Lid for Eppendorf(R) FA-45-24-11-Special Rotor
aerosol-tight, for 5424/5424R, aluminum
About This Item
UNSPSC Code:
41103913
material
aluminum
feature
aerosol-tight
manufacturer/tradename
Eppendorf® 022653067 (NA)
Eppendorf® 5424701000 (EU)
technique(s)
centrifugation: suitable
compatibility
for use with 5424 R
for use with 5424
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
Choose from one of the most recent versions:
Certificates of Analysis (COA)