Skip to Content
Merck
All Photos(7)

Key Documents

HPA027774

Sigma-Aldrich

Anti-SLPI antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-ALK1, Anti-ALP, Anti-BLPI, Anti-HUSI, Anti-HUSI-I, Anti-WAP4, Anti-WFDC4, Anti-secretory leukocyte peptidase inhibitor

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

recombinant expression
orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunohistochemistry: 1:500-1:1000

immunogen sequence

SGKSFKAGVCPPKKSAQCLRYKKPECQSDWQCPGKKRCCPDTCGIKCLDPVDTPNPTRRKPGKCPVTYGQCLMLNPPNFCEMDGQCKRDLKCCMGMCGKSCV

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SLPI(6590)

General description

Secretory leukocyte peptidase inhibitor (SLPI) is a serine-protease inhibitor which belongs to whey acidic protein family. The gene is mapped to human chromosome 20q13.12. It has four exons and encodes protein of molecular weight 11.7 kDa. It has whey-acidic-protein (WAP) motifs and the cysteine disulfide bond is crucial for its compact structure. It is secreted by epithelial cells and is present in saliva, cervical mucus and in seminal fluid.

Immunogen

secretory leukocyte peptidase inhibitor recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

Secretory leukocyte peptidase inhibitor (SLPI) functions to protect tissues during inflammation. It inhibits human immunodeficiency virus 1 (HIV-1) infection by interacting with and modulating cluster of differentiation (CD4) lymphocytes. SLPI has antimicrobial property and regulates wound healing. It also interacts with β1 tubulin in platelets and regulates proteolysis. SLPI promotes carcinogenesis in lung cancer. Low expression of SLPI in nasopharyngeal epithelium promotes Epstein-Barr virus (EBV)-mediated nasopharyngeal carcinoma (NPC). SLPI is highly expressed in ovarian and pancreatic cancer.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST86545

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide.

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Secretory leukocyte protease inhibitor promotes the tumorigenic and metastatic potential of cancer cells
Devoogdt N, et al.
Proceedings of the National Academy of Sciences of the USA, 100(10), 5778-5782 (2003)
Interactions between the megakaryocyte/platelet-specific beta1 tubulin and the secretory leukocyte protease inhibitor SLPI suggest a role for regulated proteolysis in platelet functions
Schulze H, et al.
Blood, 104(13), 3949-3957 (2004)
SLPI (secretory leukocyte peptidase inhibitor)
Ambrosi N, et al.
Atlas of Genetics and Cytogenetics in Oncology and Haematology, 20(6), 331-340 (2016)
Secretory leukocyte protease inhibitor mediates non-redundant functions necessary for normal wound healing
Ashcroft GS, et al.
Nature Medicine, 6(10), 1147-1153 (2000)
Overexpression of elafin in ovarian carcinoma is driven by genomic gains and activation of the nuclear factor kappa pathway and is associated with poor overall survival
Clauss A, et al.
Neoplasia, 12(2), IN14-IN15 (2010)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service