Skip to Content
Merck
All Photos(2)

Key Documents

HPA025690

Sigma-Aldrich

Anti-TNKS antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-PARP-5a, Anti-PARP5A, Anti-TIN1, Anti-TINF1, Anti-TNKS1, Anti-tankyrase, TRF1-interacting ankyrin-related ADP-ribose polymerase

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:20-1:50

immunogen sequence

SPDPVDGTSCCSTTSTICTVAAAPVVPAVSTSSAAGVAPNPAGSGSNNSPSSSSSPTSSSSSSPSS

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... TNKS(8658)

General description

The gene TNKS (tankyrase) is mapped to human chromosome 8p23.1. It belongs to the PARP (poly (ADP-ribose) polymerase) family of proteins and is responsible for addition of poly (ADP-ribose) (PAR) to substrate proteins. The TNKS protein contains ankyrin repeats and a sterile α-motif (SAM).

Immunogen

tankyrase, TRF1-interacting ankyrin-related ADP-ribose polymerase recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

TNKS (tankyrase) is a regulator of Wnt-signaling. It negatively regulates Axin1, thereby suppressing β-catenin degradation and causing hyperactivation of Wnt/β-catenin signaling. TNKS controls cell death and telomere length. It might also be associated with osteoarthritis. In murine lung cancer model, silencing of this gene reduces lung cancer cell growth. Application of TNKS inhibitor suppresses growth of hepatocellular carcinoma cells. Absence of TNKS activity helps in replication of human cytomegalovirus (HCMV).

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST76381

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide.

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Genetic variants in eleven telomere-associated genes and the risk of incident cardio/cerebrovascular disease: The Women's Genome Health Study.
Zee RY, et al.
Clinica Chimica Acta; International Journal of Clinical Chemistry, 412, 199-202 (2011)
A genome-wide association study of copy-number variation identifies putative loci associated with osteoarthritis in Koreans.
Moon S, et al.
BMC Musculoskelet. Disord., 16, 76-76 (2015)
Tankyrase 1 polymorphism associated with an increased risk in developing non-small cell lung cancer in a Chinese population: a proof-of-principle study.
Wang Y, et al.
International Journal of Clinical and Experimental Pathology, 8, 10500-10511 (2015)
Tankyrase inhibitors attenuate WNT/?-catenin signaling and inhibit growth of hepatocellular carcinoma cells.
Ma L, et al.
Oncotarget, 6, 25390-25401 (2015)
Sujayita Roy et al.
Viruses, 8(1) (2016-01-06)
Human cytomegalovirus (HCMV) was reported to downregulate the Wnt/β-catenin pathway. Induction of Axin1, the negative regulator of the Wnt pathway, has been reported as an important mechanism for inhibition of β-catenin. Since Tankyrase (TNKS) negatively regulates Axin1, we investigated the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service