Skip to Content
Merck
All Photos(1)

Key Documents

HPA018989

Sigma-Aldrich

Anti-CHN2 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Beta-chimaerin, Anti-Beta-chimerin, Anti-Rho GTPase-activating protein 3

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry: 1:20- 1:50

immunogen sequence

ASSNSSLSGSSVSSDAEEYQPPIWKSYLYQLQQEAPRPKRIICPREVENRPKY

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CHN2(1124)

General description

The gene CHN2 (β-chimerin) is mapped to human chromosome 7p15.3. The protein contains three domains: amino-terminal Src homology-2 (SH2) domain, the central C1 domain and a carboxyl-terminal GAP (GTPase-activating protein) domain.

Immunogen

Beta-chimaerin recombinant protein epitope signature tag (PrEST)

Application

Anti-CHN2 antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige.

Biochem/physiol Actions

CHN2 (β-chimerin) is a GTPase-activating protein (GAP) and is activated by the lipid second messenger diacylglycerol. The GAP activity of CHN2 is particularly towards Rac. Polymorphism in CHN2 is linked with diabetic retinopathy (DR) risk and non-proliferative DR risk.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST74907

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Marcelo Chen et al.
Journal of diabetes and its complications, 28(4), 460-463 (2014-05-24)
To investigate whether chimerin 2 (CHN2) genetic polymorphisms were associated with the susceptibility to diabetic retinopathy (DR) in Taiwanese individuals with type 2 diabetes. This case-control study comprised of 171 individuals with DR and 548 without DR. Four rs39059, rs2023908
Muhammad A Alvi et al.
Oncotarget, 6(25), 20863-20874 (2015-09-01)
Small bowel accounts for only 0.5% of cancer cases in the US but incidence rates have been rising at 2.4% per year over the past decade. One-third of these are adenocarcinomas but little is known about their molecular pathology and
Ameer L Elaimy et al.
Science signaling, 11(528) (2018-05-03)
The role of vascular endothelial growth factor (VEGF) signaling in cancer is not only well known in the context of angiogenesis but also important in the functional regulation of tumor cells. Autocrine VEGF signaling mediated by its co-receptors called neuropilins
Victoria Casado-Medrano et al.
Oncotarget, 7(19), 28301-28319 (2016-04-09)
β2-chimaerin is a Rac1-specific negative regulator and a candidate tumor suppressor in breast cancer but its precise function in mammary tumorigenesis in vivo is unknown. Here, we study for the first time the role of β2-chimaerin in breast cancer using
Sanghwa Yang
International journal of molecular medicine, 20(2), 225-231 (2007-07-06)
Genetic aberrations at chromosome 7 are known to be related with diverse human diseases, including cancer and autism. In a number of cancer research areas involving gastric cancer, several comparative genomic hybridization studies employing metaphase chromosome or BAC clone microarrays

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service