Skip to Content
Merck
All Photos(7)

Key Documents

HPA011144

Sigma-Aldrich

Anti-PLGRKT antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-C9orf46, Anti-Uncharacterized protein C9orf46 antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

mouse, human, rat

enhanced validation

orthogonal RNAseq
RNAi knockdown
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunohistochemistry: 1:200-1:500

immunogen sequence

KKPAFLVPIVPLSFILTYQYDLGYGTLLERMKGEAEDILETEKSKLQLPRGMITFESIEKARKEQSRFFIDK

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... C9orf46(55848)

Looking for similar products? Visit Product Comparison Guide

General description

PLGRKT (plasminogen receptor, C-terminal lysine transmembrane protein) is a transmembrane receptor, which has its C-terminal at the cell surface, containing an exposed lysine residue. It is a plasminogen receptor, and binds to it through its lysine residue. PLGRKT has a molecular weight of 17,261Da, and contains 147 amino acids. It is a highly conserved protein which shows high homology in different species. It has expression levels in catecholaminergic tissues. Its trancript is also expressed in pheochromocytoma tissues, medulla of adrenal gland, hippocampus, cerebral cortex, cerebellum and sympathetic neurons. PLGRKT is also highly expressed in human peripheral blood monocytes and monocytoid cells. This gene is present on the human chromosome 9p24.1.

Immunogen

Uncharacterized protein C9orf46.

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

PLGRKT (plasminogen receptor, C-terminal lysine transmembrane protein) is responsible for the activation of plasminogen, dependent on tissue plasminogen activator (t-PA). It binds to plasminogen through its C-terminal lysine, present on the cell surface, and interacts with t-PA, to activate plasminogen. It also co-localizes with urokinase-type plasminogen activator (uPA) receptor (uPAR), in human monocytes. PLGRKT is involved in migration, invasion, and recruitment of monocytes/macrophages in the inflammatory response, through uPA-dependent plasminogen activation. It is expressed in neuronal and neuroendocrine tissues, and might be implicated in the processes connecting plasminogen activation and catecholamine release.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST71769

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nicholas M Andronicos et al.
Blood, 115(7), 1319-1330 (2009-11-10)
Activation of plasminogen, the zymogen of the primary thrombolytic enzyme, plasmin, is markedly promoted when plasminogen is bound to cell surfaces, arming cells with the broad spectrum proteolytic activity of plasmin. In addition to its role in thrombolysis, cell surface
Hongdong Bai et al.
The Journal of biological chemistry, 286(38), 33125-33133 (2011-07-29)
Neurotransmitter release by catecholaminergic cells is negatively regulated by prohormone cleavage products formed from plasmin-mediated proteolysis. Here, we investigated the expression and subcellular localization of Plg-R(KT), a novel plasminogen receptor, and its role in catecholaminergic cell plasminogen activation and regulation
Shahrzad Lighvani et al.
Blood, 118(20), 5622-5630 (2011-09-24)
Localization of plasmin on macrophages and activation of pro-MMP-9 play key roles in macrophage recruitment in the inflammatory response. These functions are promoted by plasminogen receptors exposing C-terminal basic residues on the macrophage surface. Recently, we identified a novel transmembrane

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service