Skip to Content
Merck
All Photos(2)

Key Documents

HPA005730

Sigma-Aldrich

Anti-EPB41L2 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-41G, Anti-Band 41-like protein 2, Anti-Generally expressed protein 41

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:200-1:500

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... EPB41L2(2037)

Immunogen

Band 41-like protein 2 recombinant protein epitope signature tag (PrEST)

Sequence
ELKRNFMESTPEPRPNEWEKRRITPLSLQTQGSSHETLNIVEEKKRAEVGKDERVITEEEVEEE

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

Band 4.1-like protein 2 is a protein encoded by the EPB41L2 gene in humans. It is referred as 4.1G and 4.1-G. It is widely expressed member of the protein 4.1 family of membrane skeletal proteins. It is mapped to human chromosome 6q23 and is widely expressed among human tissues. It is known to stabilize cellular structures and proteins at the plasma membrane. The plasma membrane-associated protein regulates GPCR-mediated G(s) signaling by suppressing adenylyl cyclase-mediated cAMP production. It is found to mediate receptor/cytoskeleton interactions. The protein is known to associate with the membrane-associated guanylate kinase homologue.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST86801

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kylie J Ralston et al.
The Journal of biological chemistry, 279(32), 33816-33828 (2004-05-13)
Clustering of the T cell integrin, LFA-1, at specialized regions of intercellular contact initiates integrin-mediated adhesion and downstream signaling, events that are necessary for a successful immunological response. But how clustering is achieved and sustained is not known. Here we
Wataru Nunomura et al.
Cell biochemistry and biophysics, 66(3), 545-558 (2013-01-29)
Protein 4.1G (4.1G) is a widely expressed member of the protein 4.1 family of membrane skeletal proteins. We have previously reported that Ca(2+)-saturated calmodulin (Ca(2+)/CaM) modulates 4.1G interactions with transmembrane and membrane-associated proteins through binding to Four.one-ezrin-radixin-moesin (4.1G FERM) domain
Jeffrey M Beekman et al.
Molecular immunology, 45(7), 2069-2075 (2007-11-21)
The C-terminal domain of protein 4.1G was identified to interact with the cytosolic tail of the high affinity IgG receptor, Fc gamma RI, in yeast two-hybrid screens. Proteins of the 4.1 family have previously been found to mediate receptor/cytoskeleton interactions.
M Parra et al.
Genomics, 49(2), 298-306 (1998-05-23)
The prototypical erythrocyte membrane skeletal protein 4.1 (HGMW-approved symbol EPB41), here designated 4.1R, is encoded by a large, complexly spliced gene located on human chromosome 1p32-p33. In this paper we report evidence for a second 4.1 gene, 4.1G (HGMW-approved symbol
Toshihiro Goto et al.
Cellular signalling, 25(3), 690-697 (2012-12-04)
It has been shown lately that activity of G protein-coupled receptors (GPCRs) is regulated by an array of proteins binding to carboxy (C)-terminus of GPCRs. Proteins of 4.1 family are subsets of subcortical cytoskeletal proteins and are known to stabilize

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service