Skip to Content
Merck
All Photos(6)

Key Documents

HPA001613

Sigma-Aldrich

Anti-FBLN1 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Fibulin-1 precursor antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

recombinant expression
orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunohistochemistry: 1:50-1:200

immunogen sequence

NTLGSYLCSCSVGFRLSVDGRSCEDINECSSSPCSQECANVYGSYQCYCRRGYQLSDVDGVTCEDIDECALPTGGHICSYRCINIPGSFQCSCPSSGYRLAPNGRNCQDIDECVTGIHNCSINETCFNIQGGFRCLAFECPEN

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... FBLN1(2192)

Looking for similar products? Visit Product Comparison Guide

Immunogen

Fibulin-1 precursor recombinant protein epitope signature tag (PrEST)

Application

Anti-FBLN1 antibody produced in rabbit is suitable as an antibody suspension bead array in affinity proteomic array to explore the potential of the human proteins in plasma to predict disease status during malaria infections.
Anti-FBLN1 antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige.

Biochem/physiol Actions

FBLN1 (fibulin 1) gene encodes an extracellular matrix and plasma protein that is rich in cysteine and binds to calium. Alternative splicing of this protein produces four isoforms, A-D. It is found to be associated with fibronectin (FN). It inhibits the ability of FN to promote cell adhesion and motility by suppressing the activation of extracellular signal regulated kinase (ERK) and phosphorylation of myosin heavy chain. By inhibiting cell motility, it acts as a suppressor of tumor invasion. It serves as a potential marker for renal impairment.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST84525

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mitsuro Kanda et al.
Molecular carcinogenesis, 50(8), 571-579 (2011-01-27)
Hepatocellular carcinoma (HCC) is one of the most common cancers world-wide but the molecular mechanisms that underlie hepatocarcinogenesis are not fully determined. On the same surgical sample with HCC, we performed microarray-based gene expression profiling and karyotype analysis using a
J Qing et al.
Oncogene, 15(18), 2159-2168 (1997-12-11)
Using differential display, we identified an mRNA that is markedly down-regulated in cell line 6A/SB1, derived from a fibrosarcoma formed in an athymic mouse following injection of carcinogen-transformed MSU-1.1 cells. The nontumorigenic parental cell strain, MSU-1.1, expresses high levels of
W O Twal et al.
Journal of cell science, 114(Pt 24), 4587-4598 (2002-01-17)
Fibulin-1 is an extracellular matrix protein often associated with fibronectin (FN) in vivo. In this study, the ability of fibulin-1 to modulate adhesion, spreading and motility-promoting activities of FN was investigated. Fibulin-1 was found to have pronounced inhibitory effects on
Maja Neiman et al.
Journal of proteome research, 10(11), 4925-4934 (2011-09-06)
There is a need for reliable and sensitive biomarkers for renal impairments to detect early signs of kidney toxicity and to monitor progression of disease. Here, antibody suspension bead arrays were applied to profile plasma samples from patients with four
Y Hayashido et al.
International journal of cancer, 75(4), 654-658 (1998-02-18)
Ovarian-cancer cells are characterized by their ability to invade freely the peritoneal cavity. Estradiol stimulates the proliferation of estrogen-receptor(ER)-positive ovarian-cancer cells, as well as expression of fibulin-1, a fibronectin-binding extracellular matrix protein. Using a modified Boyden-chamber assay, we have evaluated

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service