Skip to Content
Merck
All Photos(2)

Key Documents

H6521

Sigma-Aldrich

2-Hydroxydodecanoic acid

≥98% (capillary GC)

Synonym(s):

α-Hydroxylauric acid, 2-Hydroxylauric acid, DL-α-Hydroxylauric acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)9CH(OH)COOH
CAS Number:
Molecular Weight:
216.32
EC Number:
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:
NACRES:
NA.25

Assay

≥98% (capillary GC)

lipid type

saturated FAs

storage temp.

2-8°C

SMILES string

CCCCCCCCCCC(O)C(O)=O

InChI

1S/C12H24O3/c1-2-3-4-5-6-7-8-9-10-11(13)12(14)15/h11,13H,2-10H2,1H3,(H,14,15)

InChI key

YDZIJQXINJLRLL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

DL-α-Hydroxylauric acid is a racemic mixture of D and L-α-hydroxylauric acid. DL-α-Hydroxylauric acid may be used to evaluate chiral separation methods for medium chain saturated hydroxyl fatty acids.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Vincent Sauveplane et al.
The FEBS journal, 276(3), 719-735 (2009-01-06)
An approach based on an in silico analysis predicted that CYP77A4, a cytochrome P450 that so far has no identified function, might be a fatty acid-metabolizing enzyme. CYP77A4 was heterologously expressed in a Saccharomyces cerevisiae strain (WAT11) engineered for cytochrome
Wiboonluk Pungrasmi et al.
The Journal of general and applied microbiology, 54(1), 61-69 (2008-03-08)
A bacterial strain, WL(T), which was isolated from an activated sludge, was able to degrade alkylphenols. 16S rDNA sequence analysis indicated that strain WL(T) belonged to the genus Pseudomonas (sensu stricto) and formed a monophyletic clade with the type strain
Gerald Nagahashi et al.
Fungal biology, 115(4-5), 351-358 (2011-05-03)
Two hydroxy fatty acids, tentatively identified previously in carrot root exudates, were tested for their effects on hyphal growth of the arbuscular mycorrhizal (AM) fungus, Gigaspora gigantea (Nicol. and Gerd.) Gerdemann and Trappe. Best results were achieved with a long-term bioassay
Hazel M Girvan et al.
Journal of inorganic biochemistry, 188, 18-28 (2018-08-18)
The CYP152 family of cytochrome P450 enzymes (P450s or CYPs) are bacterial peroxygenases that use hydrogen peroxide to drive hydroxylation and decarboxylation of fatty acid substrates. We have expressed and purified a novel CYP152 family member - CYP152K6 from the
Myeongjin Yi et al.
Toxicology and applied pharmacology, 330, 100-106 (2017-07-22)
Because macrophages respond to a variety of pathological and pharmacological reagents, understanding the role of P450s in macrophages is important for therapeutic intervention. There has been a lack of research on CYP4 in macrophages, but fatty acid accumulation and lipid

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service