Skip to Content
Merck
All Photos(1)

Documents

GERPN851E

ECL Plex Fluorescent Rainbow Markers

Cytiva RPN851E, pack of 500 μL

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105332
NACRES:
NA.32

feature

wetted part: no

packaging

pack of 500 μL

manufacturer/tradename

Cytiva RPN851E

color

Brownish-red

solubility

cold water: soluble (Soluble in cold water and hot water)

shipped in

dry ice

storage temp.

−20°C

Features and Benefits

  • Visible marker bands on gels and blots
  • Fluorescent images in both Cy3 and Cy5 channels

Storage and Stability

Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.

Analysis Note

To view the Certificate of Analysis for this product, please visit www.cytiva.com.

Legal Information

ECL Plex is a trademark of Cytiva
Rainbow is a trademark of Cytiva

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service