GE11-0034-93
HiTrap® MabSelect SuRe™
Cytiva 11-0034-93, pack of 5 × 1 mL
About This Item
Recommended Products
ligand
alkali-stabilized protein A-derived (E. coli)
packaging
pack of 5 × 1 mL
manufacturer/tradename
Cytiva 11-0034-93
storage condition
(20% Ehtanol)
(20% Ethanol)
parameter
flow rate
<4 ml/min flow rate
42 psi
bed size
7 mm × 25 mm
bed volume
1 mL
column I.D.
7 mm
matrix
highly cross-linked agarose
avg. part. size
85 μm (d₅₀v is the median particle size of the cumulative volume distribution)
cleaning
2-13.7
working range
3-12
capacity
≥30 mg binding capacity (humanlgG/ml at 2.4 min residence time)
suitability
suitable for bioprocess medium
storage temp.
2-8°C
Looking for similar products? Visit Product Comparison Guide
General description
HiTrap® MabSelect™ columns are prepacked with MabSelect™ a bioprocess medium for capuring of Mabs from large sample volumes.
Features and Benefits
- Withstands rigorous CIP and sanitization procedures with 0.1 to 0.5 M NaOH.
- Novel ligand design gives enhanced protease resistance resulting in lower ligand leakage.
- Oriented coupling of recombinant Protein A to the matrix via an engineered C-terminal cysteine enhances IgG binding capacity.
- Fewer regulatory concerns due to the total absence of mammalian culture in the ligand production and purification.
- Prepacked HiTrap® columns for process development, screening of purification conditions, and small-scale purification of Mabs.
Storage and Stability
Analysis Note
Other Notes
Legal Information
"MabSelect SuRe Ligand Restricted License" and "Cys-rProtein A Ligand Restricted License" are protected by the following patents and equivalent patents and patent applications in other countries: US 6,399,750 and EP 1123389. A free, non-transferable limited license to use this product for internal analytical purposes only accompanies the purchase of the product from a Cytiva company and its licensed distributors. Any other use will require a separate license from a Cytiva company.
related product
Signal Word
Warning
Hazard Statements
Precautionary Statements
Storage Class Code
3 - Flammable liquids
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Articles
Desalting at laboratory scale is a well-proven, simple, and fast method that will rapidly remove low molecular weight contaminants at the same time as transferring the sample into the desired buffer in a single step.
Protocols
This page shows how to separate IgG antibodies by affinity chromatography using MabSelect Sure from Cytiva.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service