GBL105100
Grace Bio-Labs Super G blocking buffer
volume 100 mL, sterile
Synonym(s):
blocking solution
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41161502
NACRES:
NA.25
sterility
sterile
form
liquid
packaging
pkg of 1 ea
manufacturer/tradename
Grace Bio-Labs 105100
volume
100 mL
storage temp.
2-8°C
Looking for similar products? Visit Product Comparison Guide
General description
Super G Blocking Buffer optimizes the use of nitrocellulose for protein assays using fluorescence detection. Results with Super G show significant reduction in fluorescence background while maintaining the high protein-binding capacity of porous nitrocellulose. Purified, protein-free solution has high binding efficiency to nitrocellulose for rapid and more comprehensive blocking of non-specific protein binding.
- Ultra-pure reagent
- Optimized for nitrocellulose
- Ready-to-use 1x concentration
- Protein-free solution
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service