EP951033707
Eppendorf® Deepwell Plate 96/2000 uL
sterile, volume 2000 μL, green border with clear wells, pkg of 80 plates (10 bag x 8 plates)
All Photos(1)
About This Item
UNSPSC Code:
41122107
material:
colorless wells
round bottom wells
round bottom wells
size:
96 well
sterility:
sterile
feature:
green border with clear wells
Recommended Products
material
colorless wells
round bottom wells
sterility
sterile
feature
green border with clear wells
packaging
pkg of 80 plates (10 bag x 8 plates)
manufacturer/tradename
Eppendorf® 951033707 (North America)
technique(s)
PCR: suitable
size
96 well
volume
2000 μL
working volume
2 mL
Related Categories
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
Choose from one of the most recent versions:
Certificates of Analysis (COA)