Skip to Content
Merck
All Photos(1)

Documents

EP5401000137

Eppendorf® Centrifuge 5418/5418R

5418R, refrigerated, with rotor FA-45-18-11 and rotor lid, rotary knobs, AC/DC input 120 V, 50/60 Hz, US 3-pin plug (US)

Synonym(s):

centrifuge laboratory, lab centrifuge, micro centrifuge

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
40161701

feature

CSA approved
to hold 18 1.5/2.0 mL tubes
refrigerated
rotary knobs

manufacturer/tradename

Eppendorf® 5401000137 (NA)

parameter

0-40 °C temp. range
100-14,000 rpm speed
16,873 × g max. RCF

technique(s)

centrifugation: suitable

W × H × D

21 cm × 30 cm × 21 cm

weight

22 kg

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

The Centrifuge 5418 and Centrifuge 5418 R with their 18-place capacity rotors are specifically designed for low to medium capacity in 1.5/2.0 mL tubes and microtainer. Their speed of up to 16,873 × g allows for standard molecular biology and clinical applications. Both models have an especially low access height and are characterized by a remarkably quiet operation.

Features and Benefits

  • Max. rotor capacity: 18 × 1.5/2.0 mL vessels
  • Max. speed: 16,873 × g (14,000?rpm)
  • Small footprint and low access height of 23 cm
  • OptiBowl design for quiet operation – even without a rotor lid
  • Eppendorf QuickLock” rotor allows for fast and ergonomic lid locking
  • Soft-touch one-finger lid closure for ergonomic operation
  • Full digital display with time and speed display
  • Separate short-spin key for quick centrifugation
  • Automatic lid opening at the end of the run – prevents sample warming and allows easy access to samples (only Centrifuge 5418)
  • Certified aerosol-tight rotor comes standard for safe centrifugation of hazardous samples


Special features for Centrifuge 5418 R
  • Temperature range from 0 to 40 deg. C
  • Guaranteed temperature of 4 °C at maximum rotational speed
  • FastTemp pro function for quick pre-cooling and maximum temperature accuracy inside the rotor
  • ECO power shut-off engages after 8 hours of nonuse to reduce energy consumption (46 percent over night) and extend compressor life (can be deactivated)
  • The patented compressor technology reduces vibrations and protects your samples
  • Built-in condensation drain to eliminate water accumulation in the rotor chamber

Legal Information

Eppendorf is a registered trademark of Eppendorf AG

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service