Key Documents
EP022822128
Rotor for Eppendorf® Vacufuge® Plus Concentrator
F-45-36-15, non aerosol tight, to hold, 36 x 3.0/5.0 mL flat-bottom tubes
About This Item
UNSPSC Code:
41103909
feature
to hold 36 x 15x48 mm
to hold 36 x 3.0/5.0 mL flat-bottom tubes
fixed angle rotor
non aerosol tight
packaging
pack of 1 ea
manufacturer/tradename
Eppendorf® 022822128 (NA)
technique(s)
centrifugation: suitable
compatibility
for use with centrifuge models Vacufuge Plus Concentrator
General description
Selection of rotors for the Eppendorf Vacufuge Plus.
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
Vacufuge is a registered trademark of Eppendorf AG