Skip to Content
Merck
All Photos(2)

Key Documents

EP0030624300

Eppendorf® Lobind Microplate 384/V-PP, 384 wells

white polypropylene, V-bottom clear wells, PCR clean, pkg of 80 ea

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122107
NACRES:
NB.15
material:
V-bottom clear wells
white polypropylene
size:
384 wells

material

V-bottom clear wells
white polypropylene

packaging

pkg of 80 ea

manufacturer/tradename

Eppendorf® 0030624300

technique(s)

PCR: suitable

size

384 wells

Looking for similar products? Visit Product Comparison Guide

General description

The “Eppendorf” Lobind Microplate 384/V-PP is a protein Lobind plate having 384 wells. The Lobind feature of the microplates aid in maintaining the sample quality and ensures maximum recovery of the samples. These plates are utilized in a variety of applications like sample storage and preparation.
Maximize your sample recovery with Eppendorf LoBind Plates. Eppendorf LoBind Plates are designed with a special two-component polymer mix that creates a hydrophilic surface to ensure optimal sample recovery rates. These plates are specifically designed for research or sensitive testing methods where they enable excellent results due to less sample binding to the plate surface.

Application

The “Eppendorf” Lobind Microplates are suitable for:
  • Preparation or storage of protein, peptide, or antibody samples
  • Enzymatic assays – the hydrophilic surface reduces denaturation of enzymes when they encounter the tube inner wall
  • Prevention of sample loss during storage of virus stock solutions
  • Storage of cell suspensions
  • Sample preparation in toxicology

Features and Benefits

  • The [TM=“Eppendorf”] Lobind Microplate is devoid of surface coatings to minimize sample interference.
  • Provides maximum sample recovery by significantly reducing sample binding to the surface.
  • Raised well rims and a smooth surface ensure a secure sealing in plates.
  • An optimized [TM=“RecoverMax”] well geometry to minimize the remaining volume and ensure smooth mixing properties.
  • The distinctive [TM=“OptiTrack”] matrix offers accelerated sample identification and reduces pipetting errors.
  • Free of human DNA, DNase, RNase, and PCR inhibitors (PCR clean).

Legal Information

Eppendorf is a registered trademark of Eppendorf AG

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service