Skip to Content
Merck
All Photos(1)

Key Documents

EMU027811

Sigma-Aldrich

MISSION® esiRNA

targeting mouse Usp2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

Quality Level

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

TCCTTCTGGATGGTCTCCACAATGAGGTGAACCGGGTGGCAGCAAGGCCTAAGGCCAGCCCTGAGACCCTTGATCATCTCCCTGATGAAGAAAAGGGGCGACAGATGTGGAGGAAGTATCTGGAAAGGGAAGACAGTCGGATTGGGGATCTCTTCGTTGGGCAGCTGAAGAGCTCCCTCACATGCACCGATTGTGGCTACTGCTCTACAGTCTTCGATCCCTTCTGGGATCTCTCGTTGCCCATCGCAAAGAGAGGTTACCCTGAGGTGACGTTAATGGATTGTATGAGGCTCTTCACCAAAGAGGACATATTGGATGGTGATGAGAAGCCAACTTGCTGCCGCTGCCGAGCCAGAAAACGATGCATAAAAAAGTTCTCTGTCCAGAGGTTCCCAAAGATCTTGGTGCTCCACCTGAAGCGATTCTCAGAATCCAGGATACGAACCAGCAAGCTCACAACA

Ensembl | mouse accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

B Wang et al.
Cell death and differentiation, 21(7), 1160-1169 (2014-04-29)
Mcl-1 is a unique antiapoptotic Bcl2 family member with a short half-life due to its rapid turnover through ubiquitination. We discovered that Ku70, a DNA double-strand break repair protein, functions as a deubiquitinase to stabilize Mcl-1. Ku70 knockout in mouse

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service