Skip to Content
Merck
All Photos(1)

Key Documents

EHU002981

Sigma-Aldrich

MISSION® esiRNA

targeting human KIF3A

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

Quality Level

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

GGCTGTCAGTGTGGATGAGATGAGGGGAACTATCACTGTACATAAGACTGATTCTTCCAATGAACCTCCAAAGACATTTACTTTTGATACTGTTTTTGGACCAGAGAGTAAACAACTTGATGTTTATAACTTAACTGCAAGACCTATTATTGATTCTGTACTTGAAGGCTACAATGGGACTATTTTTGCATATGGACAAACCGGAACAGGCAAAACTTTTACCATGGAAGGTGTTCGAGCTATTCCTGAACTTAGAGGAATAATTCCCAATTCATTTGCTCACATATTTGGTCATATTGCAAAAGCGGAGGGTGATACAAGATTTTTGGTTCGAGTGTCTTATTTGGAAATATATAATGAAGAAGTTCGTGACCTTTTGGGCAAGGATCAGACACAAAGGTTAGAGGTTAAAGAAAGACCTGATGTGGGAGTTTATATCAAAGATTTATCAGCTTATGTGGTAAATAATGCTGATGATATGGATAGAATTATGACGCTAGGCCACAAAAATCGTTCTGTTGGTGCAACTAATATGAACGAACATAGTTCCCGTTCCCATG

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Loren Masterson et al.
Journal of skin cancer, 2014, 596459-596459 (2014-03-19)
Due to the rarity of Merkel cell carcinoma (MCC), prospective clinical trials have not been practical. This study aimed to identify biomarkers with prognostic significance. While sixty-two patients were identified who were treated for MCC at our institution, only seventeen
Anne-Clémence Vion et al.
The Journal of cell biology, 217(5), 1651-1665 (2018-03-04)
Blood flow shapes vascular networks by orchestrating endothelial cell behavior and function. How endothelial cells read and interpret flow-derived signals is poorly understood. Here, we show that endothelial cells in the developing mouse retina form and use luminal primary cilia
Premkumar Vummidi Giridhar et al.
Journal of immunology (Baltimore, Md. : 1950), 197(11), 4228-4239 (2016-11-01)
KIF3A, the gene encoding kinesin family member 3A, is a susceptibility gene locus associated with asthma; however, mechanisms by which KIF3A might influence the pathogenesis of the disorder are unknown. In this study, we deleted the mouse Kif3a gene in
Don-Marc Franchini et al.
Cell reports, 26(1), 94-107 (2019-01-04)
Despite the clinical success of blocking inhibitory immune checkpoint receptors such as programmed cell death-1 (PD-1) in cancer, the mechanisms controlling the expression of these receptors have not been fully elucidated. Here, we identify a post-transcriptional mechanism regulating PD-1 expression

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service