CLS358037
Corning® Falcon® Cell Strainer For Use With 50ml Conical Tubes
size 40 μm, double bagged, non-sterile
Synonym(s):
culture tube
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41104932
NACRES:
NB.13
Recommended Products
material
Nylon
sterility
non-sterile
manufacturer/tradename
Corning 358037
size
40 μm
application(s)
filtration
Looking for similar products? Visit Product Comparison Guide
Related Categories
Legal Information
Corning is a registered trademark of Corning, Inc.
Falcon is a registered trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service