Skip to Content
Merck
All Photos(1)

Documents

SW 1463

90112713, human colon, Epithelial

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

product name

SW 1463, 90112713

biological source

human colon

growth mode

Adherent

karyotype

2n = 46, hypertriploid

morphology

Epithelial

products

Carcinoembryonic antigen (CEA)

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Human Caucasian rectum adenocarcinoma

Cell Line Description

Established from a grade II mucous producing and grade III solid rectal adenocarcinoma from a 66 year old Caucasian female. The cells form brush borders and produce CEA.

Application

Tumourigenicity studies

DNA Profile

STR-PCR Data: Amelogenin: X
CSF1PO: 11,12
D13S317: 12,13
D16S539: 11
D5S818: 13,14
D7S820: 9
THO1: 6,7
TPOX: 8,11
vWA: 16

Culture Medium

L15 + 2mM Glutamine + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Split subconfluent cultures 1:2 using 0.25% trypsin or trypsin/EDTA; Medium change after 4-5 days. 37°C. No CO2.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service