293T cell line human
12022001, human kidney, fetal. Small slightly rounded cells, dispersed growth
Synonym(s):
293T cell line human, 293 T, 293tsA1609neo, 293tsA1609neo Cells, HEK-293-T Cells, HEK-293T Cells, HEK293T Cells, Human Embryonic Kidney 293T Cells
About This Item
product name
293T cell line human, 12022001
biological source
human kidney (fetal)
growth mode
adherent
morphology
small slightly rounded cells growing in a dispersed manner
products
SV40 T antigen
shipped in
dry ice
storage temp.
−196°C
Looking for similar products? Visit Product Comparison Guide
General description
Cell Line Description
Application
- for lentiviral particle production
- in the expression of eukaryotic proteins
- for small interfering RNA (siRNA) transfection
- for human immunodeficiency virus (HIV) transfection for pseudoviruses production
- as a host for the transfection of pLKO.1.-TRC-ACE2 plasmid to study the localization and role of NL63-N, a nucleocapsid protein of human coronavirus NL63, in cell cycle progression
Biochem/physiol Actions
DNA Profile
Amelogenin: X
CSF1PO: 11,12
D13S317: 12
D16S539: 9,13
D5S818: 8,9
D7S820: 11
THO1: 7,9.3
TPOX: 11
vWA: 16,19
Culture Medium
Subculture Routine
Other Notes
related product
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Articles
Eukaryotic cell lines aid in complex post-translational modifications and folding of recombinant proteins. Find suitable recombinant protein production cell lines.
DNA, RNA, cDNA derived from ECACC mammalian cell lines allow screening for genes or expression patterns to identify lines most suitable for specific research.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service