Skip to Content
Merck
All Photos(1)

Key Documents

YW 62.3.20

11072202, NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA.

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

biological source

Rat x Rat Hybridoma

packaging

tube of 5 μg 11072202-DNA-5UG
pkg of vial of cells 11072202-1VL

ImmunoDon

DA rat spleen

Myeloma

Y3.AG.1.2.3

Reactivity

Anti mouse CD45 pan isoform reactive

technique(s)

cell culture | mammalian: suitable

isotype

IgG2b

shipped in

dry ice

Looking for similar products? Visit Product Comparison Guide

Cell Line Origin

Monoclonal antibody human beta 2-microglobulin

Cell Line Description

Rat hybridoma producing antibody against mouse CD45 pan isoform reactive. Cambridge Enterprise Reference number AB62:www.enterprise.cam.ac.uk.

Immunogen

CBA mouse bone marrow

Culture Medium

IDMEM (I3390) + 2 mM L-Glutamine (G7513) + 5% FBS / FCS (F2442). Where the cells are grown under static conditions, 0.02% EDTA, Trypsin (T4424) or mechanical agitation maybe required to dislodge them from the plastic.

Subculture Routine

Maintain cultures between 3-9 x 105 cells/ml. 5% CO2; 37 °C. Saturation density 9 x 105/ml.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Disclaimer

This cell line has special release conditions: Commercial organisations are required to complete the ′Cell Line Release Authorisation for Research Use in Commercial Organisations′ release conditions form.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service