ANGM-CSS
8040401, human brain, Fusiform, can form heaps of cells
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41106514
Recommended Products
product name
ANGM-CSS, 08040401
biological source
human brain
growth mode
Not specified
karyotype
Near tetraploid; range 88 to 91 - see Cell Line Description for more information
morphology
Fusiform, can form heaps of cells
products
Not specified
receptors
Not specified
technique(s)
cell culture | mammalian: suitable
relevant disease(s)
cancer
shipped in
dry ice
storage temp.
−196°C
Cell Line Origin
Human glioblastoma
Cell Line Description
A human A human glioblastoma multiforme cell line derived from a 65-year-old male. The results of immunocytochemical analysis for GFAP, Vimentin, NestiN and CD33 were positive in early and late passage cells. MGMT methylation was negative. Karyotype: Near tetraploid; range 88 to 91 with two copies of marker: der(6;7;12;)del(6q ), t(6;11), t(6;17) del(17q). Six copies of chromosome 7 and other rearrangements with loss and gain of chromosome fragments involving chromosomes 1,2,3,5,10,9,19,20. The X and Y chromosomes are present. (Information provided by Depositor)
Culture Medium
DMEM:HAMS F12 (1:1) + 2mM Glutamine + 10% Foetal Bovine Serum (FBS)
Subculture Routine
Split sub-confluent cultures (70-80%) 1:3 to 1:5 i.e. seeding at 3-4x10,000 cells/cm2 using 0.25% trypsin or trypsin/EDTA; 5% CO2;37ºC.
Other Notes
Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service