Skip to Content
Merck
All Photos(1)

Key Documents

C3710

Sigma-Aldrich

Ciliary Neurotrophic Factor human

≥95% (SDS-PAGE), recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture

Synonym(s):

CNTF

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352202
NACRES:
NA.77
Pricing and availability is not currently available.

Product Name

Ciliary Neurotrophic Factor human, CNTF, recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture

biological source

human

Quality Level

recombinant

expressed in E. coli

Assay

≥95% (SDS-PAGE)

form

lyophilized powder

potency

≤325 ng/mL ED50 ((≥ 3.1 x 103 units/mg))

quality

endotoxin tested

mol wt

22.8 kDa

packaging

pkg of 10 and 20 μg

storage condition

avoid repeated freeze/thaw cycles

technique(s)

cell culture | mammalian: suitable

impurities

≤10 EU/μg

UniProt accession no.

storage temp.

−20°C

Gene Information

General description

The CNTF (ciliary neurotrophic factor) gene encodes a cytokine,[1] and is mapped to human chromosome 11q12.1.[2]

Application

Ciliary Neurotrophic Factor human has been used to study its effect on photoreceptor neuroprotection and Muller glial cell proliferation in zebrafish retina.[1] It has also been used to study the effects of intravitreal injection of ciliary neurotrophic factor (CNTF) on photoreceptor injury and BRB (blood-retinal barrier) breakdown in transgenic model.[3]

Biochem/physiol Actions

Ciliary neurotrophic factor (CNTF) was first identified as a survival factor for neurons from the ciliary ganglion of chicken embryos. Most of its known actions are restricted to cells of the nervous system, including motor neurons, sympathetic ganglion neurons, sensory neurons, hippocampal neurons, and medial septal neurons. CNTF also prevents degeneration of motor axons after axotomy and promotes astrocyte differentiation and oligodendrocyte survival and maturation. Outside the nervous system, CNTF maintains embryonic stem cells in an undifferentiated, pluripotent state. CNTF is structurally related to leukemia inhibitory factor (LIF), interleukin-6 (IL-6), interleukin-11 (IL-11) and oncostatin M (OSM). CNTF exerts its actions through the activation of the high-affinity CTNF receptor complex, which contains the ligand-binding α-subunit (CNTF Rα) and two signal transducing β-subunits (LIF Rβ and gp130). The LIF Rβ subunit is also shared by receptors for LIF and OSM. The gp130 subunit is also shared by receptors for LIF, OSM, IL-6, and IL-11. CNTF is localized in the cell nucleus subsequent to receptor binding. Human and rat CNTF share ~83% sequence homology and show cross-reactivity in bioactivity.
Ciliary neurotrophic factor was first identified as a survival factor for neurons from the ciliary ganglion of chicken embryos. Most of its known actions are restricted to cells of the nervous system, including motor neurons, sympathetic ganglion neurons, sensory neurons, hippocampal neurons and medial septal neurons. CNTF also prevents degeneration of motor axons after axotomy and promotes astrocyte differentiation and oligodendrocyte survival and maturation.

Physical form

Lyophilized from a sterile (0.2 micron) filtered aqueous solution containing 10 mM sodium phosphate, pH 7.5

Analysis Note

The proliferative activity is tested in a cell proliferation assay using the cytokine-dependent human erythroleukemic cell line, TF-1.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

CNTF induces photoreceptor neuroprotection and Muller glial cell proliferation through two different signaling pathways in the adult zebrafish retina
Kassen SC, et al.
Experimental Eye Research, 88(6), 1051-1064 (2009)
Genomic variants, genes, and pathways of Alzheimer's disease: an overview
Naj AC, et al.
American Journal of Medical Genetics. Part B, Neuropsychiatric Genetics : the Official Publication of the International Society of Psychiatric Genetics, 174(1), 5-26 (2017)
Rachelle W Johnson et al.
Bone, 64, 47-56 (2014-04-12)
Muscle and bone are intimately linked by bi-directional signals regulating both muscle and bone cell gene expression and proliferation. It is generally accepted that muscle cells secrete factors (myokines) that influence adjacent bone cells, but these myokines are yet to
Weiyong Shen et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 32(45), 15715-15727 (2012-11-09)
Müller cells are the major glia of the retina that serve numerous functions essential to retinal homeostasis, yet the contribution of Müller glial dysfunction to retinal diseases remains largely unknown. We have developed a transgenic model using a portion of
Mansour Haidar et al.
Autophagy, 15(6), 1051-1068 (2019-01-24)
HSPB1 (heat shock protein family B [small] member 1) is a ubiquitously expressed molecular chaperone. Most mutations in HSPB1 cause axonal Charcot-Marie-Tooth neuropathy and/or distal hereditary motor neuropathy. In this study we show that mutations in HSPB1 lead to impairment

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service