Skip to Content
Merck
All Photos(1)

Documents

C2969

Sigma-Aldrich

Monoclonal Anti-I-CAM-1 (CD54) antibody produced in mouse

clone 8.4A6, purified immunoglobulin

Synonym(s):

Monoclonal Anti-CD54, Anti-I-CAM-1

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

8.4A6, monoclonal

mol wt

antigen 85-110 kDa

species reactivity

human, monkey

technique(s)

flow cytometry: suitable
immunocytochemistry: suitable
immunohistochemistry (frozen sections): suitable
indirect ELISA: suitable

isotype

IgG1

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ICAM1(3383)

Looking for similar products? Visit Product Comparison Guide

Specificity

The antibody recognizes the human CD54 (I-CAM-1) surface antigen by flow cytometry, ELISA, immunocytochemistry, and immunohistochemistry. It inhibits human leukocyte adhesion to activated endothelial cells and stimulates homotypic aggregation of SKW3 cells. The epitope recognized by the antibody is localized in domain D2 of the CD54 molecule.
5th Workshop: code no. S 100

Immunogen

TNF-α activated human endothelial cells.

Target description

CD54 (I-CAM-1) is a glycoprotein involved in mediation of intercellular adhesion. CD54 is expressed by activated endothelial cells and is also detected on cells of many other lineages and in plasma.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4.

Preparation Note

Product filtered through a 0.2 μm filter

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lanlan Tang et al.
Oncotarget, 8(16), 27314-27327 (2017-04-14)
Differentiation therapy based on all-trans-retinoic acid (ATRA) and arsenic trioxide (ATO) for the treatment of acute promyelocytic leukemia (APL) is complicated by the development of differentiation syndrome (DS), which can be fatal. We examined the role of HMGB1 (high-mobility group

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service