Skip to Content
Merck
All Photos(1)

Key Documents

AXYBF400

Breathable Rayon sealing film, for 50 μm deep well tissue culture plates

size 96 well, pkg of 10x100sheets/cs

Synonym(s):

96 multiwell plate, 96 well PCR plate, 96 well microplate, 96 well microtiter plate, 96 well plate

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122410
NACRES:
NB.22

sterility

non-sterile

form

film

manufacturer/tradename

Axygen Inc BF-400
Corning

packaging

pkg of 10x100sheets/cs

size

96 well

Looking for similar products? Visit Product Comparison Guide

General description

  • Microplate sealing films are ideal for sealing plates in various laboratory applications including PCR and Real-Time PCR to ELISA and cell culture.
  • The breathable rayon sealing films prevents sample contamination and are also suited for storage applications.
  • Chemically resistant and can withstand varying temperatures.
  • Enables efficient gas exchange for the cultivation of cellular and bacterial cultures.
  • Suited for sealing polypropylene/polystyrene culture microplates, 96-well/384-well microplates, and other assay microplates.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service