Skip to Content
Merck
All Photos(2)

Key Documents

AV31426

Sigma-Aldrich

Anti-TAF10 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-TAF10 RNA polymerase II, TATA box binding protein (TBP)-associated factor, 30 kDa

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

22 kDa

species reactivity

human, rat, bovine, mouse, guinea pig, rabbit, dog, horse

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... TAF10(6881)

General description

TAF10 is a transcription factor that forms a part of the TFIID and other regulatory complexes (such as SAGA, TFTC, STAGA and PCAF/GCN5). TAF10 is required for mouse embryogenesis and is known to establish skin barrier function in fetal mouse epidermis.
Rabbit Anti-TAF10 antibody recognizes mouse, human, canine, zebrafish, rat, bovine, and pig TAF10.

Immunogen

Synthetic peptide directed towards the C terminal region of human TAF10

Application

Rabbit Anti-TAF10 antibody can be used for western blot applications at 0.5μg/ml.

Biochem/physiol Actions

TAF10 plays a major role in transcription by binding to the TBP-associated factors (TAFs). It interacts with the AF-2-containing region E of the human estrogen receptor (ER). In conjugation with high-mobility group (HMG) protein HMG-1, TAF10 facilitates estrogen receptor-mediated transcriptional activation.

Sequence

Synthetic peptide located within the following region: DALQHCKMKGTASGSSRSKSKDRKYTLTMEDLTPALSEYGINVKKPHYFT

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Arup Kumar Indra et al.
Developmental biology, 285(1), 28-37 (2005-07-26)
TFIID, composed of the TATA box binding protein (TBP) and 13 TBP-associated factors (TAFs), plays a role in nucleating the assembly of the RNA polymerase II preinitiation complexes on protein coding genes. TAF10 (formerly TAF(II)30) is shared between TFIID and
E Scheer et al.
Genomics, 29(1), 269-272 (1995-09-01)
The basal RNA polymerase II transcription factor, TFIID, is composed of the TATA binding protein (TBP) and 8-13 TBP-associated factors (TAFs) ranging from 250 to 17 kDa. The structure of the human gene encoding the 30-kDa subunit of TFIID, TAF2H
J A Van Der Knaap et al.
The Biochemical journal, 345 Pt 3, 521-527 (2000-01-22)
The TATA-binding protein (TBP) plays a central role in eukaryotic transcription and forms protein complexes with TBP-associated factors (TAFs). The genes encoding TAF(II) proteins frequently map to chromosomal regions altered in human neoplasias. TAF(II)170 of B-TFIID is a member of
X Jacq et al.
Cell, 79(1), 107-117 (1994-10-07)
We showed previously that coactivators mediating stimulation by different activators were associated with the TATA-binding protein (TBP) in distinct TFIID complexes. We have characterized a human TBP-associated factor (TAF), hTAFII30, associated with a subset of TFIID complexes. hTAFII30 interacts with
C S Verrier et al.
Molecular endocrinology (Baltimore, Md.), 11(8), 1009-1019 (1997-07-01)
The estrogen receptor (ER) belongs to a family of ligand-inducible nuclear receptors that exert their effects by binding to cis-acting DNA elements in the regulatory region of target genes. The detailed mechanisms by which ER interacts with the estrogen response

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service