Skip to Content
Merck
All Photos(1)

Key Documents

A5595

Sigma-Aldrich

N-Acetyl-Asp-Gln-Met-Asp-7-amido-4-trifluoromethylcoumarin

≥90% (HPLC), powder

Synonym(s):

Ac-DQMD-AFC

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:

Assay

≥90% (HPLC)

form

powder

color

white

solubility

DMSO/DMF: 20 mM

storage temp.

−20°C

SMILES string

CSCC[C@H](NC(=O)[C@H](CCC(N)=O)NC(=O)[C@H](CC(O)=O)NC(C)=O)C(=O)N[C@@H](CC(O)=O)C(=O)Nc1ccc2c(OC(=O)C=C2C(F)(F)F)c1

InChI

1S/C30H35F3N6O12S/c1-13(40)35-19(11-23(42)43)29(50)37-17(5-6-22(34)41)26(47)38-18(7-8-52-2)27(48)39-20(12-24(44)45)28(49)36-14-3-4-15-16(30(31,32)33)10-25(46)51-21(15)9-14/h3-4,9-10,17-20H,5-8,11-12H2,1-2H3,(H2,34,41)(H,35,40)(H,36,49)(H,37,50)(H,38,47)(H,39,48)(H,42,43)(H,44,45)/t17-,18-,19-,20-/m0/s1

InChI key

RUWANZFFBSYLMO-MUGJNUQGSA-N

Gene Information

Amino Acid Sequence

NAc-Asp-Gln-Met-Asp-AFC

Application

Fluorogenic substrate for caspase 3 and 6.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

N J Bump et al.
Science (New York, N.Y.), 269(5232), 1885-1888 (1995-09-29)
The baculovirus antiapoptotic protein p35 inhibited the proteolytic activity of human interleukin-1 beta converting enzyme (ICE) and three of its homologs in enzymatic assays. Coexpression of p35 prevented the autoproteolytic activation of ICE from its precursor form and blocked ICE-induced
D Xue et al.
Nature, 377(6546), 248-251 (1995-09-21)
The baculovirus protein p35 inhibits programmed cell death in such diverse animals as insects, nematodes and mammals. Here we show that p35 protein is a substrate for and inhibitor of the Caenorhabditis elegans cell-death protease CED-3 (refs 6, 7) and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service