Skip to Content
Merck
All Photos(4)

Key Documents

A3010

Sigma-Aldrich

Acylase I from porcine kidney

Grade I, lyophilized powder, ≥1500 units/mg protein

Synonym(s):

Aminoacylase, N-Acylamino acid amidohydrolase

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
EC Number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.54

type

Grade I

Quality Level

form

lyophilized powder

specific activity

≥1500 units/mg protein

composition

Protein, ≥60%

UniProt accession no.

storage temp.

−20°C

Gene Information

Looking for similar products? Visit Product Comparison Guide

Application

Acylase I from porcine kidney has been used to study the acylase I-catalyzed deacetylation of various S-alkyl-N-acetyl-L-cysteines and their carbon and oxygen analogues . Acylase I may be useful to catalyze N-acetyl amino acids to enantiomerically pure L-amino acids .

Biochem/physiol Actions

Acylase I catalyzes the deacetylation of N-acetyl-L-cysteine and S-alkyl-N-acetyl-L-cysteines. n-Butylmalonic acid is an inhibitor of acylase I. S-alkyl-N-acetyl-L-cysteines with short (C0-C3) and unbranched S-alkyl substituents have been found to be good acylase I substrates .

Unit Definition

One unit will hydrolyze 1.0 μmole of N-acetyl-L-methionine per hr at pH 7.0 at 25 °C.

Analysis Note

Protein determined by biuret.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Debby Ngo et al.
JCI insight, 6(5) (2021-02-17)
Recent advances in proteomic technologies have made high-throughput profiling of low-abundance proteins in large epidemiological cohorts increasingly feasible. We investigated whether aptamer-based proteomic profiling could identify biomarkers associated with future development of type 2 diabetes (T2DM) beyond known risk factors.
K Kubo et al.
The Journal of antibiotics, 33(6), 556-565 (1980-06-01)
L-Amino acid acylase and D-amino acid acylase were stable below 50 degrees C, although the D-enzyme was more thermostable than the L-enzyme at higher temperatures. At 30 degrees C they showed the highest reaction velocity in phosphate buffer of pH
Jinit Masania et al.
Oxidative medicine and cellular longevity, 2019, 4851323-4851323 (2019-12-13)
Glycation, oxidation, nitration, and crosslinking of proteins are implicated in the pathogenic mechanisms of type 2 diabetes, cardiovascular disease, and chronic kidney disease. Related modified amino acids formed by proteolysis are excreted in urine. We quantified urinary levels of these
Y Fukagawa et al.
The Journal of antibiotics, 33(6), 543-549 (1980-06-01)
PS-5 was deacetylated to NS-5 (deacetylated PS-5) by l-amino acid acylase from porcine kidney and D-amino acid acylase from Streptomyces olivaceus but not by l-amino acid acylase from Aspergillus sp. Using PS-5, N-chloroacetyl-l-phenylalanine and N-chloroacetyl-D-valine as substrates, acylase producers were
A S Bommarius et al.
Annals of the New York Academy of Sciences, 672, 126-136 (1992-11-30)
The method of measuring enzyme deactivation by monitoring necessary addition of fresh enzyme to keep a constant degree of conversion in a CSTR at constant [E] x tau, the product of concentration of active enzyme [E] and residence time tau

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service