Skip to Content
Merck
All Photos(1)

Key Documents

113R-1

Sigma-Aldrich

CD13 (SP187) Rabbit Monoclonal Antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

100
500

conjugate

unconjugated

antibody form

culture supernatant

antibody product type

primary antibodies

clone

SP187, monoclonal

description

For In Vitro Diagnostic Use in Select Regions (See Chart)

form

buffered aqueous solution

species reactivity

human

packaging

vial of 0.1 mL concentrate (113R-14)
vial of 0.5 mL concentrate (113R-15)
bottle of 1.0 mL predilute (113R-17)
vial of 1.0 mL concentrate (113R-16)
bottle of 7.0 mL predilute (113R-18)

manufacturer/tradename

Cell Marque

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:25-1:100

isotype

IgG

control

liver

shipped in

wet ice

storage temp.

2-8°C

visualization

cytoplasmic, membranous

Gene Information

human ... ANPEP(290)

General description

CD13 (aminopeptidase-N) is a transmembrane protease present in many tissues and cell types (e.g., endothelial and epithelial cells, fibroblasts, and leukocytes). CD13 is overexpressed in various solid and hematological malignancies in humans, including acute myeloid leukemia (AML), and is thought to influence tumor progression. Acute promyelocytic leukemia, hypogranular variant, poses difficulties in morphologic interpretation for separation from other subtypes of AML. CD13 is overexpressed in the tumor and is useful in diagnosing this variant by using a panel including antibodies against CD13(+), CD34(+), CD117(+), CD16(-), and CD33(+). Myeloid sarcoma (also called chloroma, extramedullary sarcoma, or granulocytic sarcoma) is essentially a solid tumor composed of myeloblasts or immature myeloid cells in an extramedullary site. It can be isolated or occur during the course of AML, chronic myeloid leukemia, myelodysplastic syndrome, or myeloproliferative neoplasias. Myeloid sarcoma should be distinguished from large cell lymphoma, lymphoblastic lymphoma, Burkitt lymphoma, undifferentiated carcinoma, malignant melanoma, extra-medullary hematopoiesis, and inflammation. Immunochemical analyses are needed for the accurate diagnosis of myeloid sarcoma. CD13 is overexpressed in myeloid sarcoma and anti-CD13 is very useful in diagnosing myeloid sarcoma using a panel including antibodies against CD34, CD43, CD117, MPO, lysozyme, CD163 and CD68. It has been reported that CD13 is expressed in both normal and neoplastic liver tissue, where it exhibits a canalicular distribution pattern similar to that seen for polyclonal CEA and CD10. Thus, anti-CD13 can be useful as an additional marker in differentiating between HCC and non-hepatocellular neoplasms.

Quality


IVD

IVD

IVD

RUO

Linkage

CD13 Positive Control Slides, Product No. 113S, are available for immunohistochemistry (formalin-fixed, paraffin-embedded sections).

Physical form

Solution in Tris Buffer, pH 7.3-7.7, with 1% BSA and <0.1% Sodium Azide

Preparation Note

Download the IFU specific to your product lot and formatNote: This requires a keycode which can be found on your packaging or product label.

Other Notes

For Technical Service please contact: 800-665-7284 or email: service@cellmarque.com

Legal Information

Cell Marque is a trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Marion Piedfer et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 25(8), 2831-2842 (2011-05-14)
The transmembrane metalloprotease aminopeptidase-N (APN)/CD13 is overexpressed in various solid and hematological malignancies in humans, including acute myeloid leukemia (AML) and is thought to influence tumor progression. Here, we investigated the contribution of APN/CD13 to the regulation of growth and
Wojciech Gorczyca
Polish journal of pathology : official journal of the Polish Society of Pathologists, 63(1), 8-17 (2012-04-27)
A total of 97 acute promyelocytic leukemia (APL) patients with adequate flow cytometry (FC) data, bone marrow aspirates and presence of t(15;17)/PML-RARA by cytogenetics and/or FISH studies were analyzed for immunophenotypic pattern. Leukemic cells had the following phenotype: CD11b-, CD11c-
C Röcken et al.
Journal of clinical pathology, 58(10), 1069-1075 (2005-09-29)
Aminopeptidase N (CD13) is expressed in normal and neoplastic liver tissue, where it exhibits a characteristic canalicular pattern (CD13(can)), similar to that seen for CD10 and when antibodies crossreact with biliary glycoprotein I (p-CEA). To compare the putative diagnostic use
S A Pileri et al.
Leukemia, 21(2), 340-350 (2006-12-16)
Myeloid sarcoma (MS) is a rare neoplasm whose knowledge is largely based on case reports and/or technically dated contributions. Ninety-two MSs in adulthood with clinical data available were evaluated both morphologically and immunohistochemically. Seventy-four cases were also studied by fluorescent
Brigitte Bauvois et al.
Medicinal research reviews, 26(1), 88-130 (2005-10-11)
Aminopeptidase N (APN)/CD13 (EC 3.4.11.2) is a transmembrane protease present in a wide variety of human tissues and cell types (endothelial, epithelial, fibroblast, leukocyte). APN/CD13 expression is dysregulated in inflammatory diseases and in cancers (solid and hematologic tumors). APN/CD13 serves

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service