Skip to Content
Merck
All Photos(2)

Documents

242276

Sigma-Aldrich

Activated charcoal

greener alternative

DARCO®, −100 mesh particle size, powder

Synonym(s):

Charcoal activated

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C
CAS Number:
Molecular Weight:
12.01
EC Number:
MDL number:
UNSPSC Code:
11101522
PubChem Substance ID:
NACRES:
SB.52

vapor pressure

<0.1 mmHg ( 20 °C)

form

powder

autoignition temp.

842 °F

manufacturer/tradename

DARCO®

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

resistivity

1375 μΩ-cm, 20°C (graphite)

impurities

>15 g Methylene Blue adsorption (per 100 g (minimum))
<0.50% Water solubles
<12% Moisture
<200 ppm Acid soluble iron

particle size

−100 mesh

mp

3550 °C (lit.)

greener alternative category

SMILES string

[C]

InChI

1S/C

InChI key

OKTJSMMVPCPJKN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency. Click here for more details.

Application

Activated charcoal (DARCO®, -100 mesh particle size, powder) can be used in adsorption of CO2 for gas storage systems. It can be used in the formation of a composite with MIL-101, which can be used as a metal-organic framework (MOF) material for potential usage in gas separation and sensors.

Legal Information

DARCO is a registered trademark of Cabot Norit Americas, Inc.

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Supported nanometric platinum-nickel catalysts for solvent-free hydrogenation of tetralin.
Huang Y, et al.
Catalysis Communications, 69, 55-58 (2015)
Synthesis and Characterization of MIL-101 incorporated with Darco type Activated Charcoal
Panchariya DK, et al.
Materials Today, 4(2), 388-394 (2017)
Practical Alternative Synthesis of 1-(8-Fluoro-naphthalen-1-yl) piperazine.
Zhu Z, et al.
Organic Process Research & Development, 11(5), 907-909 (2007)
Measurement of CO2 adsorption kinetics on activated carbons suitable for gas storage systems
Singh VK and Kumar EA
Greenhouse Gases: Science and Technology, 7(1), 182-201 (2017)
Keywan Mortezaee et al.
Canadian journal of physiology and pharmacology, 1-12 (2015-11-18)
Melatonin is known for being beneficial in targeting liver diseases. This study aimed to investigate whether melatonin post-treatment is capable of rat carbon tetrachloride (CCl

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service