Skip to Content
Merck
All Photos(1)

Key Documents

U0020001

Ubidecarenone for system suitability

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Coenzyme Q10, Q-10, Ubiquinone 50, Ubiquinone-10

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C59H90O4
CAS Number:
Molecular Weight:
863.34
Beilstein:
1900141
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

biological source

synthetic

grade

pharmaceutical primary standard

Agency

EP

API family

ubidecarenone

form

solid

manufacturer/tradename

EDQM

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

mp

49 °C

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

O=C(C(OC)=C1OC)C(C/C=C(C)/CC/C=C(CC/C=C(CC/C=C(CC/C=C(CC/C=C(CC/C=C(C)/CC/C=C(C)/CC/C=C(C)/CCC=C(C)C)\C)\C)\C)\C)\C)=C(C)C1=O

InChI

1S/C59H90O4/c1-44(2)24-15-25-45(3)26-16-27-46(4)28-17-29-47(5)30-18-31-48(6)32-19-33-49(7)34-20-35-50(8)36-21-37-51(9)38-22-39-52(10)40-23-41-53(11)42-43-55-54(12)56(60)58(62-13)59(63-14)57(55)61/h24,26,28,30,32,34,36,38,40,42H,15-23,25,27,29,31,33,35,37,39,41,43H2,1-14H3/b45-26+,46-28+,47-30+,48-32+,49-34+,50-36+,51-38+,52-40+,53-42+

InChI key

ACTIUHUUMQJHFO-UPTCCGCDSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.
Ubidecarenone, well-known as coenzyme Q, is abundantly found in human cells, with major proportions being available in vital body organs such as heart and kidneys. It plays an important role in cell metabolism owing to its significant contribution to the electron transport pathway. It is a strong antioxidant and acts as an essential enzyme catalyst in oxidative phosphorylation taking place in the mitochondria.

Application

Ubidecarenone for system suitability may be used as an analytical reference standard for the determination of the analyte in pharmaceutical formulations by liquid chromatography.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Meyler's Side Effects of Drugs: The International Encyclopedia of Adverse Drug Reactions and Interactions
Meyler's Side Effects of Drugs: The International Encyclopedia of Adverse Drug Reactions and (2015)
Coenzyme Q10 inhibits the activation of pancreatic stellate cells through PI3K/AKT/mTOR signaling pathway
Xue R, et al.
Oncotarget, 8(54), 92300-92300 (2017)
Ubidecarenone
European Pharmacopoeia Commission and European Directorate for the Quality of Medicines & Healthcare
European pharmacopoeia, 8(54), 3875-3876 (2016)
Marcia Wyman et al.
Cleveland Clinic journal of medicine, 77(7), 435-442 (2010-07-06)
Some small clinical trials seem to show that coenzyme Q10 supplements can be used to lower blood pressure and to treat or prevent myalgia caused by hydroxymethylglutaryl coenzyme A reductase inhibitors (statins). However, larger trials are needed to determine if
Gian Paolo Littarru et al.
Nutrition (Burbank, Los Angeles County, Calif.), 26(3), 250-254 (2009-11-26)
The fundamental role of coenzyme Q(10) (CoQ(10)) in mitochondrial bioenergetics and its well-acknowledged antioxidant properties constitute the basis for its clinical applications, although some of its effects may be related to a gene induction mechanism. Cardiovascular disease is still the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service