Skip to Content
Merck
All Photos(1)

Documents

NIST93A

Borosilicate glass

NIST® SRM® 93a

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Quality Level

packaging

pkg of wafer

manufacturer/tradename

NIST®

technique(s)

X-ray spectroscopy: suitable
optical emission spectroscopy (OES): suitable

application(s)

glass & ceramic

format

matrix material

General description

The Standard Reference Material (SRM) consists of wafers measuring approximately 32 mm (1 1/4 in) in diameter and 6 mm (1/4 in) in thickness. For more information, please refer to the SDS and COA.
SRM 93A_cert

SRM 93A_SDS

Application

Borosilicate glass is intended for use in the verification of chemical analysis methods and for calibration of optical emission and x-ray spectrometric analysis methods.

Other Notes

Example analytes are listed below as a reference. Please download a current certificate at nist.gov/SRM for current analytes and certified values.
Silicon dioxide (Si02), Boric Oxide (B2O3), Sodium oxide (Na2O), Aluminium dioxide (Al2O3), Chlorine (Cl), Zirconium dioxide (ZrO2), Iron oxide (Fe2O3), Iron oxide (FeO), Potassium oxide (K2O), Titanium dioxide (TiO2), Calcium oxide (CaO), Magnesium oxide (MgO)

See certificate for values and more details at nist.gov/SRM.

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service