Skip to Content
Merck
All Photos(1)

Documents

99373

Supelco

Potassium ionophore I - cocktail B

Selectophore

Synonym(s):

B cocktail potassium ionophore

Sign Into View Organizational & Contract Pricing


About This Item

Beilstein:
78657
MDL number:
UNSPSC Code:
26111700
PubChem Substance ID:
NACRES:
NB.61

product line

Selectophore

Quality Level

form

liquid

composition

1,2-Dimethyl-3-nitrobenzene, 93.0 wt. % (40870)
Potassium ionophore I, 5.0 wt. % (60403)
Potassium tetrakis(4-chlorophenyl)borate, 2.0 wt. % (60591)

SMILES string

CC(C)[C@@H]1NC(=O)[C@H](C)OC(=O)[C@H](NC(=O)[C@H](OC(=O)[C@@H](NC(=O)[C@H](C)OC(=O)[C@H](NC(=O)[C@H](OC(=O)[C@@H](NC(=O)[C@H](C)OC(=O)[C@H](NC(=O)[C@H](OC1=O)C(C)C)C(C)C)C(C)C)C(C)C)C(C)C)C(C)C)C(C)C)C(C)C

InChI

1S/C54H90N6O18/c1-22(2)34-49(67)73-31(19)43(61)55-38(26(9)10)53(71)77-41(29(15)16)47(65)59-36(24(5)6)51(69)75-33(21)45(63)57-39(27(11)12)54(72)78-42(30(17)18)48(66)60-35(23(3)4)50(68)74-32(20)44(62)56-37(25(7)8)52(70)76-40(28(13)14)46(64)58-34/h22-42H,1-21H3,(H,55,61)(H,56,62)(H,57,63)(H,58,64)(H,59,65)(H,60,66)/t31-,32-,33-,34+,35+,36+,37-,38-,39-,40+,41+,42+/m0/s1

InChI key

FCFNRCROJUBPLU-DNDCDFAISA-N

Looking for similar products? Visit Product Comparison Guide

General description

Visit our Sensor Applications portal to learn more.

Application

Selected application examples

Legal Information

Selectophore is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Dermal - Acute Tox. 3 Oral - Aquatic Chronic 2

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

E C Reverdin et al.
Quarterly journal of experimental physiology (Cambridge, England), 71(3), 451-465 (1986-07-01)
Measurements of the K activity (aiK) in ferret ventricle were made using either single- or double-barrelled K-sensitive micro-electrodes. aiK was also estimated from the Nernst equation by measuring the membrane potential when changing the external K concentration. Micro-electrodes filled with
Neela K Codadu et al.
The Journal of physiology, 597(7), 2079-2096 (2019-01-27)
Local neocortical and hippocampal territories show different and sterotypical patterns of acutely evolving, epileptiform activity. Neocortical and entorhinal networks show tonic-clonic-like events, but the main hippocampal territories do not, unless it is relayed from the other areas. Transitions in the
Xian Zhi Fang et al.
Plant physiology, 184(4), 1900-1916 (2020-10-24)
K+ and NO3 - are the major forms of potassium and nitrogen that are absorbed by the roots of most terrestrial plants. In this study, we observed that a close relationship between NO3 - and K+ in Arabidopsis (Arabidopsis thaliana)
C J Karwoski et al.
The Journal of general physiology, 86(2), 189-213 (1985-08-01)
Recordings of light-evoked changes in extracellular K+ concentration (delta[K+]o) were obtained in the retinas of frog and mudpuppy. In eyecup preparations, various recording approaches were used and provided evidence for a K increase near the outer plexiform layer (distal K
D Ammann et al.
Neuroscience letters, 74(2), 221-226 (1987-02-24)
A valinomycin-based membrane phase for microelectrodes with relatively low electrical membrane resistances is described. Microelectrodes with tip diameters of about 1 micron exhibit resistances of about 10(10) omega. Extremely high K+ selectivities are obtained, e.g. a rejection of Na+ by

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service