Skip to Content
Merck
All Photos(1)

Key Documents

92101

Supelco

(−)-Fenchone

analytical standard

Synonym(s):

(1R)-(−)-Fenchone, (−)-1,3,3-Trimethyl-2-norbornanone, (−)-Fenchone, (1R)-1,3,3-Trimethylbicyclo[2.2.1]heptan-2-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H16O
CAS Number:
Molecular Weight:
152.23
Beilstein:
2042710
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥97.0% (GC)

optical activity

[α]/D -52.0±2.0°, neat

shelf life

limited shelf life, expiry date on the label

refractive index

n20/D 1.461 (lit.)

bp

192-194 °C (lit.)

mp

5-6 °C (lit.)

density

0.948 g/mL at 25 °C (lit.)

application(s)

food and beverages

format

neat

SMILES string

CC1(C)C2CCC(C)(C2)C1=O

InChI

1S/C10H16O/c1-9(2)7-4-5-10(3,6-7)8(9)11/h7H,4-6H2,1-3H3/t7-,10+/m0/s1

InChI key

LHXDLQBQYFFVNW-OIBJUYFYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Environment

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

151.7 °F - closed cup

Flash Point(C)

66.5 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Qi Wang et al.
eLife, 7 (2018-06-15)
Assembly, maintenance and function of synaptic junctions depend on extracellular matrix (ECM) proteins and their receptors. Here we report that Tenectin (Tnc), a Mucin-type protein with RGD motifs, is an ECM component required for the structural and functional integrity of
Yetunde Makinwa et al.
Frontiers in cell and developmental biology, 8, 813-813 (2020-09-29)
Ataxia telangiectasia and Rad3-related protein (ATR) is a serine/threonine-protein kinase of the PI3K family and is well known for its key role in regulating DNA damage responses in the nucleus. In addition to its nuclear functions, ATR also was found
Tomoya Yamaguchi et al.
Oncogene, 38(26), 5142-5157 (2019-03-22)
The receptor tyrosine kinase-like orphan receptor 1 (ROR1) is a transcriptional target of the lineage-survival oncogene NKX2-1/TTF-1 in lung adenocarcinomas. In addition to its kinase-dependent role, ROR1 functions as a scaffold protein to facilitate interaction between caveolin-1 (CAV1) and CAVIN1
Yu Matsuzawa et al.
Autophagy, 11(7), 1052-1062 (2015-06-05)
Autophagy plays important roles in metabolism, differentiation, and survival in T cells. TNFAIP3/A20 is a ubiquitin-editing enzyme that is thought to be a negative regulator of autophagy in cell lines. However, the role of TNFAIP3 in autophagy remains unclear. To
Wen Yang et al.
Cell, 167(4), 985-1000 (2016-11-25)
Mitochondrial sirtuins, SIRT3-5, are NAD+-dependent deacylases and ADP-ribosyltransferases that are critical for stress responses. However, a comprehensive understanding of sirtuin targets, regulation of sirtuin activity, and the relationships between sirtuins remains a key challenge in mitochondrial physiology. Here, we employ

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service