Skip to Content
Merck
All Photos(1)

Key Documents

87210

Sigma-Aldrich

Myristyltrimethylammonium bromide

98% (AT)

Synonym(s):

Tetradecyltrimethylammonium bromide, Trimethyl(tetradecyl)ammonium bromide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)13N(Br)(CH3)3
CAS Number:
Molecular Weight:
336.39
Beilstein:
3633227
EC Number:
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.22

description

cationic

Quality Level

Assay

≥97.5% (AT)
98% (AT)

form

crystals

mol wt

micellar avg mol wt 27,000

aggregation number

80

CMC

4-5 mM (20-25°C)

mp

245-250 °C (lit.)

solubility

H2O: 0.1 g/mL, clear

SMILES string

[Br-].CCCCCCCCCCCCCC[N+](C)(C)C

InChI

1S/C17H38N.BrH/c1-5-6-7-8-9-10-11-12-13-14-15-16-17-18(2,3)4;/h5-17H2,1-4H3;1H/q+1;/p-1

InChI key

CXRFDZFCGOPDTD-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Myristyltrimethylammonium bromide (MTAB) is a quaternary ammonium compound that consists of a long-chain alkyl group and a positively charged quaternary ammonium group. It is commonly used as a cationic surfactant and as a phase transfer catalyst in various chemical reactions.

Application

Myristyltrimethylammonium bromide can be used as a cationic surfactant to:
  • Enhance the biomass harvesting and pigments extraction from the cyanobacterium Synechocystis sp. PCC 6803.
  • Facilitate the formation of mixed micelles with amitriptyline hydrochloride.
It can also be used as a capping agent for platinum nanoparticles (Pt NPs), which enhances their catalytic activity in the CO oxidation reaction.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Irrit. 2 - STOT RE 2 Oral - STOT SE 3

Target Organs

Gastrointestinal tract, Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The role of organic capping layers of platinum nanoparticles in catalytic activity of CO oxidation
Park JY, et al.
Catalysis Letters, 129, 1-6 (2009)
How myristyltrimethylammonium bromide enhances biomass harvesting and pigments extraction from Synechocystis sp. PCC 6803
Zhou Y, et al.
Water Research, 126, 189-196 (2017)
Mixed micelle formation between amphiphilic drug amitriptyline hydrochloride and surfactants (conventional and gemini) at 293.15- 308.15 K
Rub MA,
The Journal of Physical Chemistry B, 114(19), 6354-6364 (2010)
Juan Zhang et al.
The journal of physical chemistry. B, 114(41), 13128-13135 (2010-09-28)
Two fatty acids, perfluorononanoic acid (C(8)F(17)COOH) and nonanoic acid (C(8)H(17)COOH), were mixed with a cationic hydrocarbon surfactant, tetradecyltrimethylammonium hydroxide (TTAOH), in aqueous solutions for comparative investigation. Phase behaviors of the two systems are quite different because of the special properties
Suruchi Mahajan et al.
Langmuir : the ACS journal of surfaces and colloids, 28(50), 17238-17246 (2012-12-12)
Keeping in view the use of surfactants in drug delivery, the interactions of surface active ionic liquids, such as 1-tetradecyl-3-methylimidazolium bromide (C(14)mimBr), with drugs, viz., dopamine hydrochloride (DH) and acetylcholine chloride (AC), have been studied, and the results are further

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service