Skip to Content
Merck
All Photos(1)

Documents

46317

Supelco

Metosulam

PESTANAL®, analytical standard

Synonym(s):

N-(2,6-Dichloro-3-methylphenyl)-5,7-dimethoxy-[1,2,4]triazolo[1,5-a]pyrimidine-2-sulfonamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H13Cl2N5O4S
CAS Number:
Molecular Weight:
418.26
Beilstein:
6353876
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

COc1cc(OC)n2nc(nc2n1)S(=O)(=O)Nc3c(Cl)ccc(C)c3Cl

InChI

1S/C14H13Cl2N5O4S/c1-7-4-5-8(15)12(11(7)16)20-26(22,23)14-18-13-17-9(24-2)6-10(25-3)21(13)19-14/h4-6,20H,1-3H3

InChI key

VGHPMIFEKOFHHQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazardEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2 - STOT RE 2

Target Organs

Kidney,Eyes

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C Timchalk et al.
Journal of applied toxicology : JAT, 17(1), 9-21 (1997-01-01)
This study was conducted to provide data on the pharmacokinetics of [14C]metosulam (N-[2,6-dichloro-3-methylphenyl]-5,7-dimethoxy-1,2,4-triazolo-[1,5a]- pyrimidine-2-sulfonamide). Groups of male Sprague-Dawley rats, CD-1 mice and Beagle dogs were given a single oral gavage dose of 100 mg [14C]metosulam kg-1 body weight and blood

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service