32421
Sulfaclozine sodium
VETRANAL®, analytical standard
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C10H8ClN4NaO2S
CAS Number:
Molecular Weight:
306.70
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Recommended Products
grade
analytical standard
Quality Level
product line
VETRANAL®
shelf life
limited shelf life, expiry date on the label
technique(s)
HPLC: suitable
gas chromatography (GC): suitable
application(s)
clinical testing
format
neat
SMILES string
Nc1ccc(cc1)S(=O)(=O)N([Na])c2cncc(Cl)n2
InChI
1S/C10H8ClN4O2S.Na/c11-9-5-13-6-10(14-9)15-18(16,17)8-3-1-7(12)2-4-8;/h1-6H,12H2;/q-1;+1
InChI key
NIUFFODHMKLSCH-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
Chemical structure: sulfonamide
Sulfaclozine has been used in an experimental study conducted to evaluate different persulfate activation methods.
Sulfaclozine sodium (sulfaclozine) is an anticoccidial drug.
Application
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.
Recommended products
Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.
Legal Information
VETRANAL is a registered trademark of Merck KGaA, Darmstadt, Germany
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Elimination of sulfaclozine from water with SO 4- radicals: Evaluation of different persulfate activation methods.
Ismail, Liliane, et al.
Applied Catalysis. B, Environmental, 201, 573-581 (2007)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service