Skip to Content
Merck
All Photos(1)

Key Documents

31094

Supelco

PCB No 101

analytical standard

Synonym(s):

2,2′,4,5,5′-Pentachlorobiphenyl, 2,2′,4,5,5′-PCB

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H5Cl5
CAS Number:
Molecular Weight:
326.43
Beilstein:
2507418
Ballschmiter Number:
101
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

neat

SMILES string

Clc1ccc(Cl)c(c1)-c2cc(Cl)c(Cl)cc2Cl

InChI

1S/C12H5Cl5/c13-6-1-2-9(14)7(3-6)8-4-11(16)12(17)5-10(8)15/h1-5H

InChI key

LAHWLEDBADHJGA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Polychlorinated biphenyls (PCBs) are chemical compounds used for agricultural and industrial purposes. PCBs are persistent in the environment and considered highly toxic to humans and animals.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Pictograms

Health hazardEnvironment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - STOT RE 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Xiaopeng Chen et al.
PloS one, 12(1), e0170129-e0170129 (2017-01-14)
We determined 12 polybrominated diphenyl ethers (PBDEs) and 19 polychlorinated biphenyls (PCBs) congeners in eight different brands of commercial whole milk (WM) and fat free milk (FFM) produced and distributed in California. Congeners were extracted using a modified quick, easy
Britt-Marie Bäcklin et al.
Toxicological sciences : an official journal of the Society of Toxicology, 75(1), 154-160 (2003-06-14)
Gray seal females living in the Baltic Sea have been found to exhibit a high prevalence of uterine leiomyomas. These animals are also known to accumulate lipid-soluble PCBs in their blubber. PCBs have documented endocrine-disrupting effects; to investigate whether the
Moazzam A Khan et al.
Toxicology letters, 144(2), 173-182 (2003-08-21)
Polychlorinated biphenyl compounds (PCBs) are global environmental contaminants that cause disruption of the endocrine system in humans and wildlife. Recently, we reported that acute exposures to ortho-PCB congeners 95 (2,3,6-2',5') or 101 (2,4,5,-2',5') causes changes in the performance of the
Koichi Haraguchi et al.
Chemical research in toxicology, 17(12), 1684-1691 (2004-12-21)
The tissue distributions of persistent metabolites of polychlorinated biphenyls (PCBs) in Wistar rats and homozygous uridine diphosphate glucuronosyltransferase (UGT) deficient Gunn rats exposed to 2,4,5,2',5'-pentachlorobiphenyl (CB101) and the commercial PCB mixture, Kanechlor-500 (KC500), were investigated. After exposure to CB101, four
Na He et al.
Journal of hazardous materials, 164(1), 126-132 (2008-10-01)
Pd/Fe bimetallic particles were synthesized by chemical deposition and used to dechlorinate 2,2',4,5,5'-pentachlorobiphenyl in soil. Batch experiments demonstrated that the Pd/Fe bimetallic particles could effectively dechlorinate 2,2',4,5,5'-pentachlorobiphenyl. Dechlorination was affected by several factors such as reaction time, Pd loading, the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service