Skip to Content
Merck
All Photos(1)

Key Documents

12-1540

Sigma-Aldrich

Gram′s stain solution (III)

Synonym(s):

Basic Fuchsin, Basic Violet 14, Fuchsin basic, Magenta, Rosaniline

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H20ClN3
CAS Number:
Molecular Weight:
337.85
Colour Index Number:
42510
Beilstein:
4166684
MDL number:
UNSPSC Code:
12352106

form

liquid

availability

available only in Japan

SMILES string

Cl[H].Cc1cc(ccc1N)\C(c2ccc(N)cc2)=C3\C=CC(=N)C=C3

InChI

1S/C20H19N3.ClH/c1-13-12-16(6-11-19(13)23)20(14-2-7-17(21)8-3-14)15-4-9-18(22)10-5-15;/h2-12,21H,22-23H2,1H3;1H

InChI key

NIKFYOSELWJIOF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Magenta is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service