Skip to Content
Merck
All Photos(1)

Documents

MABS2312

Sigma-Aldrich

Anti-STOM Antibody, clone GARP-50

Synonym(s):

Erythrocyte band 7 integral membrane protein;Erythrocyte membrane protein band 7.2; Protein 7.2b, Stomatin

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

GARP-50, monoclonal

mol wt

calculated mol wt 31.73 kDa
observed mol wt ~32 kDa

purified by

using protein G

species reactivity

human

species reactivity (predicted by homology)

bovine, horse, canine

packaging

antibody small pack of 100 μL

technique(s)

electron microscopy: suitable
immunodiffusion: suitable
immunofluorescence: suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG1κ

epitope sequence

N-terminal

Protein ID accession no.

UniProt accession no.

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... STOM(2040)

General description

Stomatin (UniProt: P27105; also known as Erythrocyte band 7 integral membrane protein, Erythrocyte membrane protein band 7.2, Protein 7.2b) is encoded by the STOM (also known as BND7, EPB72) gene (Gene ID: 2040) in human. Stomatin is a major integral protein of erythrocyte membranes that is palmitoylated and forms homo-oligomers and associates with lipid rafts. It contains a cytoplasmic domain (aa 1-25); an intramembrane domain (aa 26-54); and a cytoplasmic domain (aa 55-288). Its amino acid residues 265-273 are essential for homooligomerization. Stomatin was named after the rare human haemolytic anaemia hereditary stomatocytosis. It is ubiquitously expressed in all tissues and higher expression is observed in hematopoietic cells. Lower expression is observed in the brain. It is known to associate with the plasma membrane and cytoplasmic vesicles of fibroblasts, epithelial and endothelial cells, and specialized endosomes/granules in hematopoietic cells. At its N-terminal region it contains a 24-residue, basic domain that is phosphorylated at serine 10 by a cAMP-dependent protein kinase. This is followed by a 29-residue hydrophobic domain that is associated with the cytoplasmic face of the membrane and is palmitoylated at Cys-30. This hydrophobic intramembrane domain (residues 26 54) contains the conserved residue Pro-47, which is essential for its rare monotopic membrane protein structure. Adjacent to the intramembrane domain is a short sequence, KIIKEYERAII (aa 55-65) that displays similarity to the cholesterol-binding CRAC (cholesterol recognition/interaction amino acid consensus) motif. In its structure palmitoylated Cys-87 is essential for correct membrane attachment. Stomatin is known to interact with various ion channels and modulates their activity. It is also reported to inhibit the activity of Akt, suppress cancer cell proliferation, enhance apoptosis in vitro and inhibit xenograft tumor growth in vivo.Two isoforms of stomatin have been described that are produced by alternative splicing. (Ref.: Rungaldier, S., et al. (2017). PLoS One. 12(6):e0178646; Stewart, GW. (1997). Int. J. Biochem. Cell Biol. 29(2); 271-274; Christine M. Hiebi-Dirschmied, CM., et al. (1991). Biochim. Biophys. Acta. 1065; 195-202).

Specificity

Clone GARP-50 is a mouse monoclonal antibody that detects Stomatin (STOM). It targets an epitope within the N-terminal region.

Immunogen

Purified native Stomatin from human erythrocytes.

Application

Quality Control Testing

Evaluated by Western Blotting in HeLa cell lysates.

Western Blotting Analysis: A 1:500 dilution of this antibody detected STOM in HeLa cell lysates.

Tested Applications

Electron Microscopy: A representative lot detected STOM in Immunoelectron Microscopy application (Feuk-Lagerstedt, E., et al. (2002). J Leukoc Biol. 72(5):970-7).

Immunoprecipitation Analysis: A representative lot immunoprecipitated STOM in Immunoprecipitation application (Snyers, L., et al. (1998). J Biol Chem. 273(27):17221-6; Snyers, L., et al. (1999). FEBS Lett. 449(2-3):101-4).

Immunoaffinity Purification: A representative lot was used to isolate STOM by immunoaffinity chromatography..(Hiebl-Dirshmied, C.M., et al. (1991). Biochim Biophys Acta. 1065(2):195-202; Salzer, U., et al. (1993). Biochim Biophys Acta. 1151(2):149-52).

Immunofluorescence Analysis: A representative lot detected STOM in Immunofluorescence application (Snyers, L., et al. (1997). Eur J Cell Biol. 73(3):281-5; Snyers, L., et al. (1999). Eur J Cell Biol. 78(11):802-12).

Western Blotting Analysis:

Western Blotting Analysis: A representative lot detected STOM in Western Blotting application (Hiebl-Dirshmied, C.M., et al. (1991). Biochim Biophys Acta. 1065(2):195-202; Snyers, L., et al. (1998). J Biol Chem. 273(27):17221-6; Snyers, L., et al. (1999). Eur J Cell Biol. 78(11):802-12; Snyers, L., et al. (1999). FEBS Lett. 449(2-3):101-4; Salzer, U., et al. (2001). Blood. 97(4):1141-3; Mairhofer, M., et al. (2002). Blood. 100(3):897-904; Feuk-Lagerstedt, E., et al. (2002). J Leukoc Biol. 72(5):970-7).

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.

Physical form

Purified mouse monoclonal antibody IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Reconstitution

0.5 mg/mL. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Stable for 1 year at +2°C to +8°C from date of receipt.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service