Skip to Content
Merck
All Photos(2)

Key Documents

MABS1904

Sigma-Aldrich

Anti-RAMP1 Antibody, clone 4F8.1

clone 4F8.1, from mouse

Synonym(s):

Receptor activity-modifying protein 1, Calcitonin-receptor-like receptor activity-modifying protein 1, CRLR activity-modifying protein 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.43

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

4F8.1, monoclonal

species reactivity

human

packaging

antibody small pack of 25 μg

technique(s)

immunohistochemistry: suitable (paraffin)
western blot: suitable

isotype

IgG2bκ

NCBI accession no.

UniProt accession no.

target post-translational modification

unmodified

Gene Information

human ... RAMP1(10267)

Related Categories

General description

Receptor activity-modifying protein 1 (UniProt: O60894; also known as Calcitonin-receptor-like receptor activity-modifying protein 1, CRLR activity-modifying protein 1, RAMP1) is encoded by the RAMP1 gene (Gene ID: 10267) in human. RAMP1 is a single-pass type I membrane protein that is involved in the transports of calcitonin gene-related peptide type 1 receptor (CALCRL) to the plasma membrane and acts as a receptor for calcitonin-gene-related peptide (CGRP) together with CALCRL. RAMP1 lacks glycosylation sites and does not migrate to the cell surface independently. To reach the cell surface, it aggregates inside the cytoplasm and forms heterodimers with calcitonin. It is expressed in many tissues including the uterus, bladder, brain, pancreas, retina, and gastro-intestinal tract. It is synthesized with a signal peptide (aa 1-26), which is cleaved to generate the mature form. Mature RAMP1 has an extracellular domain (aa 27-117); a transmembrane domain (aa 118-138); and a short cytoplasmic tail (aa 139-148). RAMP1 is reported to acts as a vasodilation factor and mice deficient in RAMP1 exhibit inflammatory responses with a transient, but significant, increase in serum calcitonin-gene-related peptide levels and pro-inflammatory cytokines. (Ref.: Nakayama, T., et al. (2017). J. Hum. Hypertens. 31(5); 361-365).

Specificity

Clone 4F8.1 specifically detects Receptor activity-modifying protein 1 (RAMP1) in human cells. It targets an epitope within 91 amino acids from the internal region.

Immunogen

GST/His-tagged recombinant fragment corresponding to 91 amino acids from the internal region of human Receptor activity-modifying protein 1 (RAMP1).

Application

Anti-RAMP1, clone 4F8.1, Cat. No. MABS1904, is a mouse monoclonal antibody that detects Receptor activity-modifying protein 1 (RAMP1) and has been tested for use in Immunohistochemistry (Paraffin) and Western Blotting.
Immunohistochemistry Analysis: A 1:250 dilution from a representative lot detected RAMP1 in human uterus and human colon tissue sections.
Research Category
Signaling

Quality

Evaluated by Western Blotting in human brain tissue lysate.

Western Blotting Analysis: 0.5 µg/mL of this antibody detected RAMP1 in human brain tissue lysate.

Target description

~17 kDa observed; 16.99 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Format: Purified
Protein G purified
Purified mouse monoclonal antibody IgG2b in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Camille Cohen et al.
Nature communications, 15(1), 743-743 (2024-01-26)
Chronic kidney disease (CKD) is a public health problem driven by myofibroblast accumulation, leading to interstitial fibrosis. Heterogeneity is a recently recognized characteristic in kidney fibroblasts in CKD, but the role of different populations is still unclear. Here, we characterize

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service