MABN453
Anti-CHRDL1 Antibody, clone 10C9.3
clone 10C9.3, from mouse
Synonym(s):
Chordin-like protein 1, Neuralin-1, Neurogenesin-1, Ventroptin
About This Item
biological source
mouse
Quality Level
antibody form
purified immunoglobulin
antibody product type
primary antibodies
clone
10C9.3, monoclonal
species reactivity
human, rat
technique(s)
immunohistochemistry: suitable
western blot: suitable
isotype
IgG1κ
NCBI accession no.
UniProt accession no.
shipped in
wet ice
target post-translational modification
unmodified
Gene Information
human ... CHRDL1(91851)
General description
Immunogen
Application
Neuroscience
Quality
Western Blotting Analysis: 1.0 µg/mL of this antibody detected CHRDL1 in 200 µg of rat brain tissue lysate.
Target description
Physical form
Storage and Stability
Analysis Note
Western Blotting Analysis: 1.0 µg/mL from a representative lot detected CHRDL1 in 200 µg of human brain tissue lysate.
Immunohistochemistry Analysis: A 1:50-1,000 dilution from a representative lot detected CHRDL1 in human retina, human cerebellum, rat cerebellum, and rat cerebral cortex tissue.
Other Notes
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service