MABN1564
Anti-DYRK1A Antibody, clone 11D9.1
clone 11D9.1, from mouse
Synonym(s):
Dual specificity tyrosine-phosphorylation-regulated kinase 1A, EC 2.7.12.1, Dual specificity YAK1-related kinase, HP86, Protein kinase minibrain homolog, MNBH, hMNB
About This Item
biological source
mouse
antibody form
purified immunoglobulin
antibody product type
primary antibodies
clone
11D9.1, monoclonal
species reactivity
human
packaging
antibody small pack of 25 μg
technique(s)
western blot: suitable
isotype
IgG2bκ
NCBI accession no.
UniProt accession no.
target post-translational modification
unmodified
Gene Information
human ... DYRK1A(1859)
Related Categories
General description
Specificity
Immunogen
Application
Quality
Western Blotting Analysis: 1 µg/mL of this antibody detected DYRK1A in human cerebral cortex tissue lysate.
Target description
Physical form
Other Notes
Not finding the right product?
Try our Product Selector Tool.
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service