Skip to Content
Merck
All Photos(1)

Documents

MABF2085

Sigma-Aldrich

Anti-CD11b/CD18 Antibody, clone mAb107

clone mAb107, from mouse

Synonym(s):

Integrin alpha-M/ITGB2, CR3

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.43

biological source

mouse

antibody form

purified antibody

antibody product type

primary antibodies

clone

mAb107, monoclonal

species reactivity

nonhuman primates, human

packaging

antibody small pack of 25 μg

technique(s)

dot blot: suitable
flow cytometry: suitable
immunohistochemistry: suitable
inhibition assay: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

target post-translational modification

unmodified

Gene Information

human ... ITGAM(3684)

General description

Integrin alpha-M (UniProt: P11215; also known as CD11 antigen-like family member B, CR-3 alpha chain, Cell surface glycoprotein MAC-1 subunit alpha, Leukocyte adhesion receptor MO1, Neutrophil adherence receptor, CD11b) is encoded by the ITGAM (also known as CD11B, CR3A) gene (Gene ID: 3684) in human. Integrins are alpha/beta heterodimeric adhesion receptors that couple the extracellular matrix on other cells with the contractile cytoskeleton to transduce mechanochemical signals across the plasma membrane. CD11b is a single-pass type I membrane protein that is synthesized with a signal peptide (aa 1-16), which is cleaved off in the mature form. It is predominantly expressed in monocytes and granulocytes and some expression is also observed in neutrophils. It is usually found as a heterodimer of alpha and beta subunits and associates with Integrin beta-2 (ITGB2). Two isoforms of CD11b have been reported that are produced by alternative splicing. CD11b contains a large extracellular domain (aa 17-1104), a short transmembrane domain (aa 1105-1128), and a cytoplasmic domain (aa 1129-1152). It has been implicated in various adhesive interactions of monocytes, macrophages and granulocytes as well as in mediating the uptake of complement-coated particles. It contains a metal ion-dependent adhesion site (MIDAS) in the integrin A domain, which is essential for stabilization of domain A in high affinity state. The major ligand recognition site of CD11b/CD18 has been mapped to the A-domain in the CD11b subunit (CD11bA). Both magnesium and calcium ions exhibit equal affinities for the inactive CD11bA, however, upon activation there is a 10-fold increase in the binding affinity of magnesium ions without affecting affinity of calcium ions. Mutations in ITGAM gene are known to cause Systemic lupus erythematosus 6, which is characterized by wide range of system dysfunctions, principally involving skin, joints, kidneys and serosal membranes. (Ref.: Zerria, K., et al. (2006). Immunology 119(4); 431-440; Mahalingam, B., et al., (2011). J. Immunol. 187 (12); 6393-6401; Li, R., et al. (2002). J. Immunol. 168 (3); 1219-1225.).

Specificity

Clone mAb 107 detects CD11b/CD18 in human neutrophils. It targets an epitope with in the A domain.

Immunogen

GST-tagged recombinant fragment corresponding to 211 amino acids from the N-terminal half of human CD11bA.

Application

Anti-CD11b/CD18, clone mAb107, Cat. No. MABF2085, is a mouse monoclonal antibody that detects CD11b/CD18 nd has been tested for use in Dot Blot, Flow Cytometry, Immunohistochemistry, and Inhibition assay.
Dot Blot Analysis: A representative lot detected CD11b/CD18 in Dot Blot applications (Li, R., et. al. (2002). J Immunol. 168(3):1219-25).

Flow Cytometry Analysis: A representative lot detected CD11b/CD18 in Flow Cytometry applications (Mahalingam, B., et. al. (2011). J Immunol. 187(12):6393-401; Li, R., et. al. (2002). J Immunol. 168(3):1219-25).

Inhibition Analaysis: A representative lot prophylactically inactivated CD11b/CD18 and prevents CD11b/CD18 binding to multiple ligands (Dehnadi, A., et. al. (2017). Nat Commun. 8:13899; Li, R., et. al. (2002). J Immunol. 168(3):1219-25).

Immunohistochemistry Analysis: A representative lot detected CD11b/CD18 in Immunohistochemistry applications (Dehnadi, A., et. al. (2017). Nat Commun. 8:13899).
Research Category
Cell Structure

Quality

Evaluated by Flow Cytometry in human neutrophils.

Flow Cytometry Analysis: 1 µg of this antibody detected CD11b/CD18 in one million human neutrophils.

Target description

127.18 kDa calculated.

Physical form

Format: Purified
Protein G purified
Purified mouse monoclonal antibody IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at -20°C from date of receipt. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service